Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03957734_s1
          See other 5830426I08RIK GE Assays ›
          Gene Symbol:
          5830426I08Rik
          Gene Name:
          RIKEN cDNA 5830426I08 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03957734_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 5830426I08Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK020019.1 - 1 252 74
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AV314912.1 - 1 159 Chr. 15 - 25225502 - 25225764 [-] on Build GRCm38 74
          GenBankmRNA BY717835.1 - 1 Chr. 15 - 25225490 - 25225854 [-] on Build GRCm38 74

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 2824 Gm2824 -197.8kb Sense Chr. 15 - 24978422 - 25027708 on NCBI Build GRCm38
          Downstream RIKEN cDNA 4930445E18 gene 4930445E18Rik 55.6kb Sense Chr. 15 - 25281439 - 25332648 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12337063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12344026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11849816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12077966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12424666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079779 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12077957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12077965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12337058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30079801 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106100164 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b7b59cd5b-z8v7b:80/100.66.130.62:80.
          git-commit: 87deb889a5bb48f859ebd9970e87be562fd9396c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0