Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03958446_s1
          See other 9430024F10RIK GE Assays ›
          Gene Symbol:
          9430024F10Rik
          Gene Name:
          RIKEN cDNA 9430024F10 gene
          Gene Aliases:
          B930059L03Rik
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03958446_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9430024F10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK020436.1 - 1 946 112
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB367970.2 - 1 397 Chr. 12 - 110590550 - 110591033 [-] on Build GRCm38 112
          GenBankmRNA BB089922.1 - 1 184 Chr. 12 - 110590513 - 110590818 [-] on Build GRCm38 112

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream protein phosphatase 2,
          regulatory subunit B',
          gamma
          Ppp2r5c 2610043M05Rik,
          2700063L20Rik,
          AI060890,
          AW545884,
          C85228,
          D12Bwg0916e,
          mKIAA0044
          -7.4kb Anti-sense Chr. 12 - 110447179 - 110583075 on NCBI Build GRCm38
          Downstream dynein cytoplasmic 1 heavy chain 1 Dync1h1 9930018I23Rik,
          AI894280,
          DHC1,
          DHC1a,
          DNCL,
          Dnchc1,
          Dnec1,
          Dnecl,
          Loa,
          MAP1C,
          P22,
          Swl,
          mKIAA0325
          9.8kb Anti-sense Chr. 12 - 110601395 - 110666944 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032553 AK020436.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032553 AK020436.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1430730_PM_at - -
          Mouse Genome 430 2.0 Array 1444000_at - -
          Mouse Genome 430 2.0 Array 1430730_at - -
          Mouse Genome 430 2.0 Array 1444000_PM_at - -
          Mouse Genome 430 2.0 Array 1430730_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77271_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77271_at - -
          Mouse Expression Set 430 1430730_at - -
          Mouse Expression Set 430 1444000_at - -
          Mouse Expression Set 430 1430730_PM_at - -
          Mouse Expression Set 430 1444000_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444000_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430730_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444000_at - -
          Affymetrix HT Mouse Genome 430B Array 1430730_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 716017 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0