Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03958529_s1
          See other C430049B03RIK GE Assays ›
          Gene Symbol:
          C430049B03Rik
          Gene Name:
          RIKEN cDNA C430049B03 gene
          Gene Aliases:
          2700063P19Rik, 9430052C07Rik, mir-322, mir-351, mir-503
          Chromosome Location:
          Chr.X: 53053112 - 53057190 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03958529_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C430049B03Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038184.1 4 2877 83
          NR_038185.1 4 2591 83
          GenBank mRNA AK020467.1 - 1 50 83
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CN839732.1 - 1 365 Chr. X - 53053114 - 53053592 [-] on Build GRCm38 83
          GenBankmRNA BY721752.1 - 1 48 Chr. X - 53053112 - 53053275 [-] on Build GRCm38 83

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 351 Mir351 Mirn351,
          mmu-mir-351
          Sense Chr. X - 53053255 - 53053353 on NCBI Build GRCm38
          Downstream microRNA 503 Mir503 Mirn503,
          mmu-mir-503
          708.0 Sense Chr. X - 53053984 - 53054054 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00002234.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977910 AK012483.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114652 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127812 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00014152.mm.1 TC0X00002234.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00014152.mm.1 NR_029776.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00014152.mm.1 NR_029776.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00014152.mm.1 NR_038184.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00014152.mm.1 NR_038184.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00014152.mm.1 NR_038185.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00014152.mm.1 NR_038185.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00014151.mm.1 TC0X00002234.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00014151.mm.1 NR_038184.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00014151.mm.1 NR_038184.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00014151.mm.1 NR_038185.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00014151.mm.1 NR_038185.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 396251 - -
          ABI Mouse Genome Survey Microarray 531323 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 102640433 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245433 - -
          Illumina Sentrix mouse-6.v1 bead array 102640433 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102640433 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2546861 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0