Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03959121_s1
          See other A930007D18RIK GE Assays ›
          Gene Symbol:
          A930007D18Rik
          Gene Name:
          RIKEN cDNA A930007D18 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03959121_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A930007D18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK020824.1 - 1 1055 94
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB277748.1 - 1 162 Chr. 4 - 83472737 - 83473020 [+] on Build GRCm38 94
          GenBankmRNA BE133797.1 - 1 Chr. 4 - 83472472 - 83473019 [-] on Build GRCm38 94
          GenBankmRNA CD352192.1 - 1 Chr. 4 - 83472336 - 83473081 [-] on Build GRCm38 94
          GenBankmRNA BB283846.1 - 1 397 Chr. 4 - 83472499 - 83473020 [+] on Build GRCm38 94
          GenBankmRNA BI965727.1 - 1 Chr. 4 - 83472548 - 83473012 [-] on Build GRCm38 94
          GenBankmRNA AW909438.1 - 1 Chr. 4 - 83472566 - 83473019 [-] on Build GRCm38 94
          GenBankmRNA CX212678.1 - 1 406 Chr. 4 - 83472493 - 83473019 [+] on Build GRCm38 94
          GenBankmRNA CF582058.1 - 1 Chr. 4 - 83472243 - 83473013 [-] on Build GRCm38 94

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream PC4 and SFRS1 interacting protein 1 Psip1 AA408851,
          AU015605,
          Dfs70,
          Ledgfa,
          Ledgfb,
          Psip2
          Anti-sense Chr. 4 - 83455680 - 83486638 on NCBI Build GRCm38
          Downstream coiled-coil domain containing 171 Ccdc171 4930418J05Rik,
          4930473A06Rik,
          A330015D16Rik
          52.5kb Sense Chr. 4 - 83525537 - 83866143 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2295821 AK020824.1 mouse_transcriptome
          Agilent Mouse cDNA Microarray (G4104A) 991550 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1429890_PM_at - -
          Mouse Genome 430 2.0 Array 1429890_PM_at - -
          Mouse Genome 430 2.0 Array 1429890_at - -
          Mouse Expression Set 430 1429890_at - -
          Mouse Expression Set 430 1429890_PM_at - -
          Murine Genome U74v2 Set 135583_at - -
          Murine Genome U74v2 Set 135583_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429890_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429890_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 438458 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104230692 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0