Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03970428_g1
          See other GM13293 GE Assays ›
          Gene Symbol:
          Gm13293
          Gene Name:
          predicted gene 13293
          Gene Aliases:
          OTTMUSG00000011311
          Chromosome Location:
          Chr.2: 11339489 - 11344106 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03970428_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm13293
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040369.1 3 - 4 659 79
          GenBank mRNA AK140161.1 - 3 - 4 659 79

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream protein kinase C,
          theta
          Prkcq A130035A12Rik,
          AW494342,
          PKC-0,
          PKC-theta,
          PKCtheta,
          Pkcq
          -41.2kb Sense Chr. 2 - 11172382 - 11301226 on NCBI Build GRCm38
          Downstream 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3 Pfkfb3 E330010H22Rik,
          iPFK-2,
          uPFK-2
          127.3kb Anti-sense Chr. 2 - 11471430 - 11554071 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000215.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC1700002078.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_01_of_03_v2 A_68_P01424230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02047176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21005655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21005672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21005658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21005647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21005657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21005661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01464450 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01464447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01464446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02047177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01424231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21005648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01464448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01424238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01518355 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200000798.mm.1 TC0200000215.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200000798.mm.1 NR_040369.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000798.mm.1 NR_040369.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000799.mm.1 TC0200000215.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200000799.mm.1 NR_040369.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000799.mm.1 NR_040369.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0