Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03979563_m1
          See other GM20337 GE Assays ›
          Gene Symbol:
          Gm20337
          Gene Name:
          predicted gene, 20337
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 80942652 - 80945399 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03979563_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm20337
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045057.1 1 - 2 665 63
          GenBank mRNA AK145430.1 - 1 - 2 664 63
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY756110.1 - 1 - 2 227 Chr. 12 - 80942673 - 80944916 [-] on Build GRCm38 63
          GenBankmRNA BG069753.2 - 2 - 3 Chr. 12 - 80942652 - 80944789 [+] on Build GRCm38 63
          GenBankmRNA BQ963819.1 - 1 - 2 628 Chr. 12 - 80942857 - 80945374 [-] on Build GRCm38 63

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 1700052I22 gene 1700052I22Rik -18.6kb Sense Chr. 12 - 80923432 - 80924447 on NCBI Build GRCm38
          Downstream serine/arginine-rich splicing factor 5 Srsf5 Sfrs5 106.0 Anti-sense Chr. 12 - 80945504 - 80950507 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200002010.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 259299 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 774691 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116182 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163225 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163225 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116182 - -
          Whole Mouse Genome G4122A A_52_P519008 AK145430.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116182 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163225 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200013819.mm.1 TC1200002010.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200013819.mm.1 NR_045057.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013819.mm.1 NR_045057.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013820.mm.1 TC1200002010.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200013820.mm.1 NR_045057.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013820.mm.1 NR_045057.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013821.mm.1 TC1200002010.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200013821.mm.1 NR_045057.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013821.mm.1 NR_045057.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013822.mm.1 TC1200002010.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200013822.mm.1 NR_045057.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013822.mm.1 NR_045057.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0