Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03983082_s1
          See other GM19589 GE Assays ›
          Gene Symbol:
          Gm19589
          Gene Name:
          predicted gene, 19589
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 88551517 - 88562166 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03983082_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm19589
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_037971.1 6 2360 84
          GenBank mRNA AK136671.1 - 2 1452 84
          BB818892.1 - 1 83

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream glutamine repeat protein 1 Glrp1 GRP-1 -50.5kb Anti-sense Chr. 1 - 88499871 - 88510066 on NCBI Build GRCm38
          Downstream ADP-ribosylation factor-like 4C Arl4c A630084M22Rik,
          Arl7,
          LAK
          136.1kb Anti-sense Chr. 1 - 88698225 - 88702191 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100000849.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01025805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00000129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00235456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424406 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00190834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00629853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00291456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00371973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00371955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00985696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01037507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00371980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00585910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00492796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01002809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00000128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00492806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00371976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00814860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00181020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01037509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00492804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424385 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00585908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20424372 - -
          Array Name Probe IDs Targets Target Type
          Mouse Genome 430 2.0 Array 1444710_PM_at - -
          Mouse Genome 430 2.0 Array 1444710_at - -
          Affymetrix Mouse Clariom S Array PSR0100007293.mm.1 TC0100000849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100007293.mm.1 NR_037971.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100007293.mm.1 NR_037971.1 Refseq
          Mouse Expression Set 430 1444710_at - -
          Mouse Expression Set 430 1444710_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 504137 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6bbccd986b-mx52t:80/100.66.130.62:80.
          git-commit: 717389b07c2b9f75290ff02e811d41bb86d4254a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0