Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000012833.mm.1 |
TC1000001761.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012833.mm.1 |
NM_001163590.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012833.mm.1 |
NM_001163590.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012833.mm.1 |
NM_001163591.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012833.mm.1 |
NM_001163591.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012833.mm.1 |
NM_029075.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012833.mm.1 |
NM_029075.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012832.mm.1 |
TC1000001761.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012832.mm.1 |
NM_001163590.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012832.mm.1 |
NM_001163590.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012832.mm.1 |
NM_001163591.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012832.mm.1 |
NM_001163591.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012832.mm.1 |
NM_029075.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012832.mm.1 |
NM_029075.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012838.mm.1 |
TC1000001761.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012838.mm.1 |
NM_001163590.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012838.mm.1 |
NM_001163590.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012838.mm.1 |
NM_001163591.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012838.mm.1 |
NM_001163591.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012838.mm.1 |
NM_029075.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012838.mm.1 |
NM_029075.1 |
Refseq |