Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03991737_m1
          See other 9430038I01RIK GE Assays ›
          Gene Symbol:
          9430038I01Rik
          Gene Name:
          RIKEN cDNA 9430038I01 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 137375569 - 137411900 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03991737_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9430038I01Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029886.2 NP_084162.1 3 - 4 342 70
          GenBank mRNA AK020460.1 - 3 - 4 314 70
          AK078159.1 - 3 - 4 322 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004465.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174371 AK051763.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174371 AK171836.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2116833 AV013327.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2116833 NM_029886.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2116833 AI413306.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537880 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 330828 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 536179 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 188967 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 284852 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 942789 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 252054 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 319565 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 140857 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 194018 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 407069 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 359072 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 473539 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 130047 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 616217 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 432730 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 187943 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 291236 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 616488 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 825488 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 79774 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 495098 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 321459 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 21539 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 119720 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 102792 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 21523 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 100364 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143030 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179904 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2116833 AV013327.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2116833 NM_029886.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2116833 AI413306.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174371 AK051763.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174371 AK171836.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06386886 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143030 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179904 - -
          Whole Mouse Genome G4122A A_51_P236588 AK020460.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P236588 AK051763.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P236588 AK078159.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P236588 AK171836.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P236588 NM_029886.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179904 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06289199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06191103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06152357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06297076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06503625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05900395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05916225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06476131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06296475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05973775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06109263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06108584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05916224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05900404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05916226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06108570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25565763 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10568759 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568760 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568761 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568762 - -
          Affymetrix HT MG-430 PM Array Plate 1435159_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1431198_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1453799_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10568758 - -
          Mouse Genome 430 2.0 Array 1431198_x_at - -
          Mouse Genome 430 2.0 Array 1435159_PM_at - -
          Mouse Genome 430 2.0 Array 1431198_PM_x_at - -
          Mouse Genome 430 2.0 Array 1453799_at - -
          Mouse Genome 430 2.0 Array 1453799_PM_at - -
          Mouse Genome 430 2.0 Array 1435159_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77252_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77252_at - -
          Affymetrix Mouse Clariom S Array PSR0700037460.mm.1 TC0700004465.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037460.mm.1 NM_029886.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037460.mm.1 NM_029886.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037461.mm.1 TC0700004465.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037461.mm.1 NM_029886.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037461.mm.1 NM_029886.2 Refseq
          Mouse Expression Set 430 1453799_PM_at - -
          Mouse Expression Set 430 1435159_PM_at - -
          Mouse Expression Set 430 1431198_PM_x_at - -
          Mouse Expression Set 430 1453799_at - -
          Mouse Expression Set 430 1431198_x_at - -
          Mouse Expression Set 430 1435159_at - -
          Murine Genome U74v2 Set 131002_PM_at - -
          Murine Genome U74v2 Set 131002_at - -
          Affymetrix HT Mouse Genome 430B Array 1431198_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1435159_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453799_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431198_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1435159_at - -
          Affymetrix HT Mouse Genome 430B Array 1453799_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 385508 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 100780059 - -
          Illumina mouse-6 v1.0 expression beadchip 102030397 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2601254 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2576671 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2601254 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253929 - -
          Illumina Sentrix mouse-6.v1 bead array 102030397 - -
          Illumina Sentrix mouse-6.v1 bead array 100780059 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102030397 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100780059 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1233136 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1253929 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6bbccd986b-mx52t:80/100.66.130.62:80.
          git-commit: 717389b07c2b9f75290ff02e811d41bb86d4254a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0