Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03991852_g1
          See other TMEM161A GE Assays ›
          Gene Symbol:
          Tmem161a
          Gene Name:
          transmembrane protein 161A
          Gene Aliases:
          AI428876, BB161850, BC021367
          Chromosome Location:
          Chr.8: 70172329 - 70183681 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03991852_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem161a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001293797.1 NP_001280726.1 10 - 11 1118 109
          NM_145597.4 NP_663572.1 11 - 12 1222 109
          NR_122028.1 10 - 11 1127 109
          GenBank mRNA AK032711.1 - 7 - 8 738 109
          AK049979.1 - 11 - 12 1221 109
          AK082916.1 - 10 - 11 1119 109
          AK147962.1 - 11 - 12 1215 109
          AK167517.1 - 5 - 6 525 109
          AK212182.1 - 1 - 2 110
          AK212718.1 - 1 - 2 51 109
          BC021367.1 - 11 - 12 1222 5030434 109
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AA874611.1 - 3 - 4 336 Chr. 8 - 70181003 - 70182078 [+] on Build GRCm38 109
          GenBankmRNA CK129602.1 - 3 - 4 367 Chr. 8 - 70180961 - 70182294 [+] on Build GRCm38 109
          GenBankmRNA AA815743.1 - 2 - 3 186 Chr. 8 - 70181239 - 70182009 [+] on Build GRCm38 109
          GenBankmRNA BQ177703.1 - 2 - 3 281 Chr. 8 - 70181146 - 70182380 [+] on Build GRCm38 109
          GenBankmRNA CX220965.1 - 2 - 3 153 Chr. 8 - 70181274 - 70182429 [+] on Build GRCm38 109
          GenBankmRNA CF551360.1 - 1 - 2 106 Chr. 8 - 70181429 - 70182569 [+] on Build GRCm38 109

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream myocyte enhancer factor 2B Mef2b AI451606 -13.9kb Sense Chr. 8 - 70152778 - 70167488 on NCBI Build GRCm38
          Downstream solute carrier family 25,
          member 42
          Slc25a42 2900084M01Rik 659.0 Anti-sense Chr. 8 - 70184340 - 70212281 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem161a

          Gene Ontology Categories:

          Function(s) Process(es)

          response to retinoic acid
          cellular response to oxidative stress
          cellular response to UV
          positive regulation of DNA repair
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
          molecular_function

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline