Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03992571_s1
          See other APOL11B GE Assays ›
          Gene Symbol:
          Apol11b
          Gene Name:
          apolipoprotein L 11b
          Gene Aliases:
          A330102K04Rik
          Chromosome Location:
          Chr.15: 77633951 - 77643286 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03992571_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apol11b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001143686.1 NP_001137158.1 4 1849 83
          GenBank mRNA AK039744.1 - 3 1728 83
          AY662646.1 - 5 2083 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001817.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453736 AY662646.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453736 NM_001143686.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453736 AK039744.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453736 AY662646.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453736 NM_001143686.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P453736 AK039744.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 367039 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 361568 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 87356 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453736 AY662646.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453736 NM_001143686.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453736 AK039744.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453736 AY662646.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453736 NM_001143686.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P453736 AK039744.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03075700 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03075706 - -
          Whole Mouse Genome G4122A A_51_P453736 AY662646.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453736 NM_001143686.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453736 AK039744.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453736 AY662646.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453736 NM_001143686.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453736 AK039744.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03075700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30354987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12354032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30354995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12338461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30354998 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10430194 - -
          Affymetrix HT MG-430 PM Array Plate 1457307_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1440927_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10430190 - -
          Mouse Genome 430 2.0 Array 1457307_at - -
          Mouse Genome 430 2.0 Array 1440927_x_at - -
          Mouse Genome 430 2.0 Array 1440927_PM_x_at - -
          Mouse Genome 430 2.0 Array 1457307_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 328563_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 328563_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500014093.mm.1 TC1500001817.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500014093.mm.1 NM_001143686.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014093.mm.1 NM_001143686.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014095.mm.1 TC1500001817.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500014095.mm.1 NM_001143686.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014095.mm.1 NM_001143686.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014090.mm.1 TC1500001817.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500014090.mm.1 NM_001143686.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014090.mm.1 NM_001143686.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014091.mm.1 TC1500001817.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500014091.mm.1 NM_001143686.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014091.mm.1 NM_001143686.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014094.mm.1 TC1500001817.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500014094.mm.1 NM_001143686.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014094.mm.1 NM_001143686.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014099.mm.1 TC1500001817.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500014099.mm.1 NM_001143686.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014099.mm.1 NM_001143686.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014092.mm.1 TC1500001817.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500014092.mm.1 NM_001143686.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014092.mm.1 NM_001143686.1 Refseq
          Mouse Expression Set 430 1457307_at - -
          Mouse Expression Set 430 1440927_x_at - -
          Mouse Expression Set 430 1440927_PM_x_at - -
          Mouse Expression Set 430 1457307_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440927_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1457307_at - -
          Affymetrix HT Mouse Genome 430B Array 1440927_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1457307_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 563253 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1252157 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252157 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0