Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04180279_mH
          See other SERPINB9G GE Assays ›
          Gene Symbol:
          Serpinb9g Serpinb9f
          Gene Name:
          serine (or cysteine) peptidase inhibitor, clade B, member 9g
          serine (or cysteine) peptidase inhibitor, clade B, member 9f
          Gene Aliases:
          1600002F03Rik, AW545779, NK21B
          BC052216, NK21, NK21L1, Spi13, ovalbumin
          Chromosome Location:
          Chr.13: 33484790 - 33496002 on Build GRCm38
          Chromosome Location:
          Chr.13: 33324077 - 33335370 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm04180279_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpinb9g
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011455.3 NP_035585.2 5 - 6 647 104
          GenBank mRNA AF425083.1 - 5 - 6 642 104
          AK005394.1 - 5 - 6 647 104
          BC064758.1 - 5 - 6 632 30018322 104
          BC064759.1 - 5 - 6 622 30432705 104
          Gene Symbol: Serpinb9f
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183197.1 NP_899020.1 5 - 6 636 104
          GenBank mRNA BC052216.1 - 5 - 6 636 30020424 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000369.mm.1, TC1300000371.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serpinb9g
          Click here to find siRNAs for Gene Serpinb9f

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of endopeptidase activity
          hematopoietic progenitor cell differentiation
          serine-type endopeptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971425 NM_183197.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971425 BC052216.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971425 NM_183197.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971425 BC052216.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P28933718 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28933731 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10404477 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404467 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404474 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404473 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404485 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404480 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404468 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404483 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404475 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404481 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404465 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404478 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404469 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404482 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404470 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404471 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404484 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404466 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404476 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404486 - -
          Affymetrix Mouse Gene 1.1 ST Array 10404479 - -
          Affymetrix Mouse Gene 1.1 ST Array 10404464 - -
          Affymetrix Mouse Gene 1.1 ST Array 10404472 - -
          Affymetrix Mouse Clariom S Array PSR1300002222.mm.1 TC1300000369.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002222.mm.1 NM_183197.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002222.mm.1 NM_183197.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002244.mm.1 TC1300000371.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002244.mm.1 NM_011455.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002244.mm.1 NM_011455.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002243.mm.1 TC1300000371.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002243.mm.1 NM_011455.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002243.mm.1 NM_011455.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002221.mm.1 TC1300000369.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002221.mm.1 NM_183197.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002221.mm.1 NM_183197.1 Refseq
          Mouse Expression Set 430 1418423_s_at - -
          Mouse Expression Set 430 1418423_PM_s_at - -
          Murine Genome U74v2 Set 161115_PM_r_at - -
          Murine Genome U74v2 Set 161115_r_at - -
          Mouse Genome 430A 2.0 Array 1418423_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1418423_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2493397 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2493397 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0