Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04206200_m1
          See other RBP7 GE Assays ›
          Gene Symbol:
          Rbp7
          Gene Name:
          retinol binding protein 7, cellular
          Gene Aliases:
          1110002J23Rik, CRBP-III
          Chromosome Location:
          Chr.4: 149449687 - 149466462 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04206200_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rbp7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022020.2 NP_071303.1 3 - 4 372 98
          GenBank mRNA AF260923.1 - 3 - 4 373 98
          AK003307.1 - 2 - 3 180 98
          AY141971.1 - 3 - 4 382 98
          BC028432.1 - 3 - 4 369 1348253 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400004041.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rbp7

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          transporter activity
          retinoid binding
          lipid binding
          retinal binding
          retinol binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23400564 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23400566 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23400567 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23400563 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23400565 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343627 AY141971.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343627 BC028432.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343627 AK003307.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343627 NM_022020.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343627 AF260923.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343627 AY141971.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343627 BC028432.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343627 AK003307.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343627 NM_022020.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343627 AF260923.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 636399 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 252717 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 78973 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 92293 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101784 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343627 AY141971.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343627 BC028432.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343627 AK003307.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343627 NM_022020.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343627 AF260923.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343627 AY141971.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343627 BC028432.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343627 AK003307.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343627 NM_022020.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343627 AF260923.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P508899 AY141971.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P508899 BC028432.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P508899 NM_022020.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P508899 AF260923.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23400622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23400596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23400625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23400623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23400619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23400607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23400593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23400627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23400602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23400628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23400629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23400599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23400621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23400604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23400609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23400612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23400606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23400624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23400601 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101784 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01015770 - -
          Whole Mouse Genome G4122A A_51_P508899 AY141971.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P508899 BC028432.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P508899 NM_022020.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P508899 AF260923.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343627 AY141971.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343627 BC028432.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343627 AK003307.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343627 NM_022020.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343627 AF260923.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343627 AY141971.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343627 BC028432.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343627 AK003307.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343627 NM_022020.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343627 AF260923.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130546 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101784 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03318447 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03318450 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03318451 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03839084 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23400565 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23400564 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23400566 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23400563 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23400567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23400567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400571 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03639475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400598 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03987063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04032485 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03476441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03985185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400572 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03370433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400604 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03562098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400609 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03819458 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03708308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400640 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03309409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400601 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23400588 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10518675 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518677 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518678 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518676 - -
          Affymetrix HT MG-430 PM Array Plate 1449461_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10518674 - -
          Mouse Genome 430 2.0 Array 1449461_PM_at - -
          Mouse Genome 430 2.0 Array 1449461_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 63954_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 63954_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400033572.mm.1 TC0400004041.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400033572.mm.1 NM_022020.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033572.mm.1 NM_022020.2 Refseq
          Mouse Expression Set 430 1449461_at - -
          Mouse Expression Set 430 1449461_PM_at - -
          Mouse Genome 430A 2.0 Array 1449461_at - -
          Mouse Genome 430A 2.0 Array 1449461_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449461_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449461_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 928859 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1340369 - -
          MouseWG-6 v2 Expression BeadChip 1740286 - -
          Illumina mouse-6 v1.0 expression beadchip 1740286 - -
          Illumina mouse-6 v1.0 expression beadchip 1340369 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001340369 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001740286 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1218914 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2733708 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218914 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2733708 - -
          mouse-6 v1.1 (Illumina) scl0002608.1_109-S - -
          mouse-6 v1.1 (Illumina) scl23496.4.1_61-S - -
          Illumina Sentrix mouse-6.v1 bead array 1740286 - -
          Illumina Sentrix mouse-6.v1 bead array 1340369 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002608.1_109-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl23496.4.1_61-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1340369 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1740286 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2733708 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1218914 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1218914 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2733708 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0