Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04206620_m1
          See other FAS GE Assays ›
          Gene Symbol:
          Fas
          Gene Name:
          Fas (TNF receptor superfamily member 6)
          Gene Aliases:
          AI196731, APO1, APT1, CD95, TNFR6, Tnfrsf6, lpr
          Chromosome Location:
          Chr.19: 34290659 - 34327775 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04206620_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fas
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001146708.1 NP_001140180.1 1 - 2 86 110
          NM_007987.2 NP_032013.2 1 - 2 86 110
          GenBank mRNA AK086933.1 - 1 - 2 63 110
          BC061160.1 - 1 - 2 59 30302649 110
          BY348234.1 - 1 - 2 87 110
          M83649.1 - 1 - 2 79 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000497.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK086933.1, BC061160.1, M83649.1, NM_001146708.1, NM_007987.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fas

          Gene Ontology Categories:

          Function(s) Process(es)

          immunoglobulin production
          renal system process
          apoptotic process
          activation-induced cell death of T cells
          inflammatory cell apoptotic process
          immune response
          signal transduction
          extrinsic apoptotic signaling pathway via death domain receptors
          response to toxic substance
          gene expression
          regulation of gene expression
          positive regulation of cell death
          B cell mediated immunity
          dendrite regeneration
          positive regulation of protein homooligomerization
          tumor necrosis factor-mediated signaling pathway
          regulation of cell population proliferation
          regulation of apoptotic process
          T cell homeostasis
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative thymic T cell selection
          regulation of lymphocyte differentiation
          regulation of myeloid cell differentiation
          spleen development
          negative regulation of B cell activation
          protein homooligomerization
          response to glucocorticoid
          neuron apoptotic process
          cellular response to lithium ion
          cellular response to hyperoxia
          positive regulation of release of cytochrome c from mitochondria
          motor neuron apoptotic process
          apoptotic signaling pathway
          extrinsic apoptotic signaling pathway
          extrinsic apoptotic signaling pathway in absence of ligand
          hepatocyte apoptotic process
          necroptotic signaling pathway
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          protease binding
          transmembrane signaling receptor activity
          tumor necrosis factor-activated receptor activity
          protein binding
          kinase binding
          identical protein binding
          protein-containing complex binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1900004437.mm.1 TC1900000497.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900004437.mm.1 NM_001146708.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004437.mm.1 NM_001146708.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004437.mm.1 NM_007987.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004437.mm.1 NM_007987.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004436.mm.1 TC1900000497.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900004436.mm.1 NM_001146708.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004436.mm.1 NM_001146708.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004436.mm.1 NM_007987.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004436.mm.1 NM_007987.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004438.mm.1 TC1900000497.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900004438.mm.1 NM_001146708.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004438.mm.1 NM_001146708.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004438.mm.1 NM_007987.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004438.mm.1 NM_007987.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 857676 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0