Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04206922_mH
          See other GM14548 GE Assays ›
          Gene Symbol:
          Gm14548 Pira11 Pira6 Pirb
          Gene Name:
          predicted gene 14548
          paired-Ig-like receptor A11
          paired-Ig-like receptor A6
          paired Ig-like receptor B
          Gene Aliases:
          OTTMUSG00000017169
          Gm14548, p91C
          12M1, Pira10, p91B
          Gp91, LIR-3, Lilrb3, PIR-B
          Chromosome Location:
          Chr.7: 3884201 - 3898159 on Build GRCm38
          Chromosome Location:
          -
          Chromosome Location:
          Chr.7: 4274124 - 4284827 on Build GRCm38
          Chromosome Location:
          Chr.7: 3712505 - 3720456 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          Assay ID Mm04206922_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm14548
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001166672.1 NP_001160144.1 4 - 5 666 102
          Gene Symbol: Pira11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011088.1 NP_035218.1 667 102
          GenBank mRNA AF041036.1 - 658 102
          AF055896.1 - 658 102
          BC103528.2 - 667 40039374 102
          BC103529.1 - 667 40039371 102
          Gene Symbol: Pira6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289428.1 NP_001276357.1 723 102
          NM_008848.1 NP_032874.1 658 102
          NM_011093.1 NP_035223.1 704 102
          GenBank mRNA AF041035.1 - 658 102
          U96687.1 - 704 102
          Gene Symbol: Pirb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011095.2 NP_035225.2 4 - 5 714 102
          GenBank mRNA AF148878.1 - 660 102
          AF148879.1 - 669 102
          AF148880.1 - 4 - 5 665 102
          AF288356.1 - 664 102
          AK142375.1 - 4 - 5 686 102
          AK152232.1 - 4 - 5 690 102
          AK153116.1 - 4 - 5 690 102
          AK161081.1 - 685 102
          AK165255.1 - 4 - 5 714 102
          AK170618.1 - 3 - 4 841 102
          AK170720.1 - 3 - 4 849 102
          BC026937.1 - 668 4488338 102
          U83172.1 - 4 - 5 658 102

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gm14548
          Click here to find siRNAs for Gene Pira11
          Click here to find siRNAs for Gene Pira6
          Click here to find siRNAs for Gene Pirb

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          B cell homeostasis
          immune system process
          cytokine-mediated signaling pathway
          B cell mediated immunity
          myeloid dendritic cell differentiation
          molecular_function

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0