Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04206923_gH
          See other GM14548 GE Assays ›
          Gene Symbol:
          Gm14548 Pira11 Pira2 Pira4 Pira7 Pirb
          Gene Name:
          predicted gene 14548
          paired-Ig-like receptor A11
          paired-Ig-like receptor A2
          paired-Ig-like receptor A4
          paired-Ig-like receptor A7
          paired Ig-like receptor B
          Gene Aliases:
          OTTMUSG00000017169
          Gm14548, p91C
          6M23
          6M7
          16M2
          Gp91, LIR-3, Lilrb3, PIR-B
          Chromosome Location:
          Chr.7: 3884201 - 3898159 on Build GRCm38
          Chromosome Location:
          -
          Chromosome Location:
          Chr.7: 3836810 - 3845099 on Build GRCm38
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          Chr.7: 3712505 - 3720456 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          Assay ID Mm04206923_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm14548
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001166672.1 NP_001160144.1 5 - 6 950 82
          Gene Symbol: Pira11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF055896.1 - 942 82
          Gene Symbol: Pira2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011089.2 NP_035219.2 5 - 6 970 82
          GenBank mRNA AK137475.1 - 5 - 6 971 82
          AK140776.1 - 5 - 6 970 82
          Gene Symbol: Pira4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011091.1 NP_035221.1 950 82
          GenBank mRNA U96685.1 - 950 82
          Gene Symbol: Pira7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011094.1 NP_035224.1 5 - 6 978 82
          GenBank mRNA U96688.1 - 5 - 6 978 82
          Gene Symbol: Pirb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011095.2 NP_035225.2 5 - 6 998 82
          GenBank mRNA AF148880.1 - 5 - 6 949 82
          AK152232.1 - 5 - 6 974 82
          AK153116.1 - 5 - 6 974 82
          AK161081.1 - 969 82
          AK165255.1 - 5 - 6 998 82
          AK170618.1 - 4 - 5 1125 82
          AK170720.1 - 4 - 5 1133 82
          U83172.1 - 5 - 6 942 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004631.mm.1, TC0700004634.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gm14548
          Click here to find siRNAs for Gene Pira11
          Click here to find siRNAs for Gene Pira2
          Click here to find siRNAs for Gene Pira4
          Click here to find siRNAs for Gene Pira7
          Click here to find siRNAs for Gene Pirb

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          B cell homeostasis
          immune system process
          cytokine-mediated signaling pathway
          B cell mediated immunity
          myeloid dendritic cell differentiation
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700019369.mm.1 TC0700004634.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019369.mm.1 NM_011089.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019369.mm.1 NM_011089.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019310.mm.1 TC0700004631.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019310.mm.1 NM_011095.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019310.mm.1 NM_011095.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019386.mm.1 TC0700004634.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019386.mm.1 NM_001166672.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019386.mm.1 NM_001166672.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0