Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700019305.mm.1 |
TC0700004631.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700019305.mm.1 |
NM_011095.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019305.mm.1 |
NM_011095.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019383.mm.1 |
TC0700004634.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700019383.mm.1 |
NM_001166672.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019383.mm.1 |
NM_001166672.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019384.mm.1 |
TC0700004634.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700019384.mm.1 |
NM_001166672.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019384.mm.1 |
NM_001166672.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000402.mm.1 |
TC0700000030.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700000402.mm.1 |
NM_008848.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000402.mm.1 |
NM_008848.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000401.mm.1 |
TC0700000030.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700000401.mm.1 |
NM_008848.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000401.mm.1 |
NM_008848.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000401.mm.1 |
NM_011093.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000401.mm.1 |
NM_011093.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019367.mm.1 |
TC0700004634.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700019367.mm.1 |
NM_011089.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019367.mm.1 |
NM_011089.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019381.mm.1 |
TC0700004634.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700019381.mm.1 |
NM_001166672.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019381.mm.1 |
NM_001166672.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019306.mm.1 |
TC0700004631.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700019306.mm.1 |
NM_011095.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019306.mm.1 |
NM_011095.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019382.mm.1 |
TC0700004634.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700019382.mm.1 |
NM_001166672.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019382.mm.1 |
NM_001166672.1 |
Refseq |
Mouse Expression Set 430 |
1420464_s_at |
- |
- |
Mouse Expression Set 430 |
1420464_PM_s_at |
- |
- |
Murine Genome U74v2 Set |
100328_s_at |
- |
- |
Murine Genome U74v2 Set |
100328_PM_s_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1420464_PM_s_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1420464_s_at |
- |
- |