Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04207183_m1
          See other SERPINA3A GE Assays ›
          Gene Symbol:
          Serpina3a
          Gene Name:
          serine (or cysteine) peptidase inhibitor, clade A, member 3A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 104106704 - 104121897 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04207183_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpina3a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001167705.1 NP_001161177.1 2 - 3 857 98
          GenBank mRNA BC063325.1 - 2 - 3 1047 30280343 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001038.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of peptidase activity
          negative regulation of endopeptidase activity
          serine-type endopeptidase inhibitor activity
          peptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2161456 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2161455 BC063325.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2161455 NM_001167705.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117525 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2161456 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2161455 BC063325.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2161455 NM_001167705.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117525 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02594893 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07733813 - -
          Whole Mouse Genome G4122A A_51_P125779 NM_001167705.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P125779 AK016702.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105867 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10190589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28701521 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10398027 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398030 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398029 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398028 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398026 - -
          Affymetrix HT MG-430 PM Array Plate 1421556_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10398025 - -
          Mouse Genome 430 2.0 Array 1421556_PM_at - -
          Mouse Genome 430 2.0 Array 1421556_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74069_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74069_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1200007392.mm.1 TC1200001038.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200007392.mm.1 NM_001167705.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007392.mm.1 NM_001167705.1 Refseq
          Mouse Expression Set 430 1421556_PM_at - -
          Mouse Expression Set 430 1421556_at - -
          Mouse Genome 430A 2.0 Array 1421556_at - -
          Mouse Genome 430A 2.0 Array 1421556_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421556_at - -
          Affymetrix HT Mouse Genome 430A Array 1421556_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 668691 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6020088 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2610378 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2610378 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0