Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019647.6 | NP_062621.2 | 2 - 3 | 364 | 80 | |
GenBank mRNA | AK002940.1 | - | 2 - 3 | 111 | 80 | |
AK008457.1 | - | 2 - 3 | 239 | 80 | ||
AK018480.1 | - | 2 - 3 | 110 | 80 | ||
AK146440.1 | - | 2 - 3 | 105 | 80 | ||
AK168998.1 | - | 2 - 3 | 455 | 80 | ||
BC086904.1 | - | 2 - 3 | 111 | 6763320 | 80 | |
BC086905.1 | - | 2 - 3 | 109 | 6775274 | 80 | |
BC086935.1 | - | 2 - 3 | 109 | 6764354 | 80 | |
BC089582.1 | - | 2 - 3 | 110 | 6751295 | 80 | |
BC090256.1 | - | 96 | 4024411 | 80 | ||
BC096039.1 | - | 1 - 2 | 94 | 80 | ||
BC096404.1 | - | 2 - 3 | 105 | 1331663 | 80 | |
BC106116.1 | - | 2 - 3 | 100 | 3985115 | 80 | |
BC138299.1 | - | 2 - 3 | 99 | 8861318 | 80 | |
BC138300.1 | - | 2 - 3 | 99 | 8861319 | 80 | |
U93863.1 | - | 1 - 2 | 106 | 80 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |