Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sphk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172472.1 | NP_001165943.1 | 3 - 4 | 603 | 102 | |
NM_001172473.1 | NP_001165944.1 | 3 - 4 | 489 | 102 | ||
NM_001172475.1 | NP_001165946.1 | 3 - 4 | 528 | 102 | ||
NM_011451.3 | NP_035581.1 | 1 - 2 | 260 | 102 | ||
NM_025367.6 | NP_079643.2 | 3 - 4 | 486 | 102 | ||
GenBank mRNA | AF068748.1 | - | 3 - 4 | 528 | 102 | |
AF068749.1 | - | 1 - 2 | 260 | 102 | ||
AF415213.1 | - | 1 - 2 | 158 | 102 | ||
AK028667.1 | - | 1 - 2 | 2550 | 102 | ||
AK136289.1 | - | 2 - 3 | 274 | 102 | ||
AK155332.1 | - | 3 - 4 | 588 | 102 | ||
AK160900.1 | - | 3 - 4 | 479 | 102 | ||
BC037710.1 | - | 3 - 4 | 401 | 4221357 | 102 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |