Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04207354_m1
          See other CRYGA GE Assays ›
          Gene Symbol:
          Cryga
          Gene Name:
          crystallin, gamma A
          Gene Aliases:
          Cryg-4, DGcry-4
          Chromosome Location:
          Chr.1: 65100389 - 65103363 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04207354_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cryga
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007774.3 NP_031800.1 2 - 3 282 101
          GenBank mRNA BC056430.1 - 2 - 3 269 6824353 101
          BY150722.1 - 2 - 3 293 101
          CF728682.1 - 2 - 3 282 101
          CF738355.1 - 2 - 3 282 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003880.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cryga

          Gene Ontology Categories:

          Function(s) Process(es)

          eye development
          lens development in camera-type eye
          structural constituent of eye lens

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P508156 CF738355.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P508156 NM_007774.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P508156 BC056430.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P508156 CF728682.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P508156 CF738355.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P508156 NM_007774.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P508156 BC056430.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P508156 CF728682.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 639989 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115334 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P508156 CF738355.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P508156 NM_007774.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P508156 BC056430.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P508156 CF728682.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P508156 CF738355.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P508156 NM_007774.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P508156 BC056430.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P508156 CF728682.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P508156 CF738355.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P508156 NM_007774.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P508156 BC056430.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P508156 CF728682.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P508156 CF738355.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P508156 NM_007774.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P508156 BC056430.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P508156 CF728682.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296406 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296410 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296405 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296403 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296420 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296390 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296396 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115334 - -
          Whole Mouse Genome G4122A A_51_P508156 CF738355.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P508156 NM_007774.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P508156 BC056430.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P508156 CF728682.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P508156 CF738355.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P508156 NM_007774.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P508156 BC056430.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P508156 CF728682.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116357 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115334 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00015184 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00015185 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00015179 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00156850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00156850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296423 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296406 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296424 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00015179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296385 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00015181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296392 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296427 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00781477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00156847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296417 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296416 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00156849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296421 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10355204 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355203 - -
          Affymetrix HT MG-430 PM Array Plate 1454333_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1422652_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10355202 - -
          Mouse Genome 430 2.0 Array 1454333_at - -
          Mouse Genome 430 2.0 Array 1454333_PM_at - -
          Mouse Genome 430 2.0 Array 1422652_PM_at - -
          Mouse Genome 430 2.0 Array 1422652_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12964_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12964_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100031607.mm.1 TC0100003880.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100031607.mm.1 NM_007774.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100031607.mm.1 NM_007774.3 Refseq
          Mouse Expression Set 430 1422652_at - -
          Mouse Expression Set 430 1422652_PM_at - -
          Mouse Expression Set 430 1454333_PM_at - -
          Mouse Expression Set 430 1454333_at - -
          Murine Genome U74v2 Set 103609_at - -
          Murine Genome U74v2 Set 103609_PM_at - -
          Murine Genome U74v2 Set 169050_r_at - -
          Murine Genome U74v2 Set 169050_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1422652_PM_at - -
          Mouse Genome 430A 2.0 Array 1422652_at - -
          Affymetrix HT Mouse Genome 430A Array 1422652_at - -
          Affymetrix HT Mouse Genome 430A Array 1422652_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454333_at - -
          Affymetrix HT Mouse Genome 430B Array 1454333_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 850560 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2340309 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002340309 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2705692 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2705692 - -
          mouse-6 v1.1 (Illumina) scl012964.1_261-S - -
          Illumina Sentrix mouse-6.v1 bead array 2340309 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl012964.1_261-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2340309 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2705692 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2705692 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0