Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04207507_gH
          See other IFNA11 GE Assays ›
          Gene Symbol:
          Ifna11 Ifna2
          Gene Name:
          interferon alpha 11
          interferon alpha 2
          Gene Aliases:
          IFN-[a]11
          Ifa2
          Chromosome Location:
          Chr.4: 88819919 - 88820565 on Build GRCm38
          Chromosome Location:
          Chr.4: 88683207 - 88683779 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm04207507_gH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ifna11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008333.2 NP_032359.2 1 351 115
          GenBank mRNA BC116870.1 - 1 351 40126189 115
          BC116872.1 - 1 351 40126191 115
          Gene Symbol: Ifna2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC100688.1 - 1 309 115
          BC100689.1 - 1 310 115
          BC100698.1 - 1 310 115
          BC100699.1 - 1 310 115
          BC114423.1 - 1 304 115
          K01411.1 - 1 149 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000905.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ifna11
          Click here to find siRNAs for Gene Ifna2

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          T cell activation involved in immune response
          natural killer cell activation involved in immune response
          defense response
          humoral immune response
          cytokine-mediated signaling pathway
          B cell differentiation
          positive regulation of peptidyl-serine phosphorylation of STAT protein
          B cell proliferation
          response to exogenous dsRNA
          defense response to virus
          cytokine activity
          cytokine receptor binding
          type I interferon receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2036475 NM_008333.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2036475 NM_008333.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P157288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092358 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23092364 - -
          Whole Mouse Genome G4122A A_51_P157288 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127522 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132511 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23092336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03528738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03430993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03758551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03899094 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10514316 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505882 - -
          Affymetrix Mouse Gene 1.1 ST Array 10514315 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505881 - -
          Mouse Genome 430 2.0 Array 1422332_PM_at - -
          Mouse Genome 430 2.0 Array 1422332_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15964_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15964_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400006097.mm.1 TC0400000905.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400006097.mm.1 NM_008333.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006097.mm.1 NM_008333.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006099.mm.1 TC0400000905.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400006099.mm.1 NM_008333.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006099.mm.1 NM_008333.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006098.mm.1 TC0400000905.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400006098.mm.1 NM_008333.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006098.mm.1 NM_008333.2 Refseq
          Mouse Expression Set 430 1422332_PM_at - -
          Mouse Expression Set 430 1422332_at - -
          Murine Genome U74v2 Set 100671_at - -
          Murine Genome U74v2 Set 96419_PM_f_at - -
          Murine Genome U74v2 Set 100671_PM_at - -
          Murine Genome U74v2 Set 101788_PM_f_at - -
          Murine Genome U74v2 Set 96419_f_at - -
          Murine Genome U74v2 Set 101788_f_at - -
          Mouse Genome 430A 2.0 Array 1422332_PM_at - -
          Mouse Genome 430A 2.0 Array 1422332_at - -
          Affymetrix HT Mouse Genome 430A Array 1422332_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422332_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 718990 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103850711 - -
          Illumina mouse-6 v1.0 expression beadchip 103850711 - -
          Illumina mouse-6 v1.0 expression beadchip 1570097 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001570097 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2769241 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252609 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2769241 - -
          mouse-6 v1.1 (Illumina) scl015964.1_328-S - -
          mouse-6 v1.1 (Illumina) GI_6680364-S - -
          Illumina Sentrix mouse-6.v1 bead array 1570097 - -
          Illumina Sentrix mouse-6.v1 bead array 103850711 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl015964.1_328-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1570097 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103850711 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2769241 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2769241 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0