Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04207573_g1
          See other MCM7 GE Assays ›
          Gene Symbol:
          Mcm7
          Gene Name:
          minichromosome maintenance deficient 7 (S. cerevisiae)
          Gene Aliases:
          AI747533, Mcmd7, mCDC47
          Chromosome Location:
          Chr.5: 138164589 - 138171862 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04207573_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mcm7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008568.2 NP_032594.1 2 - 3 215 87
          GenBank mRNA AK132748.1 - 776 87
          AK133091.1 - 2 - 3 211 87
          AK144980.1 - 2 - 3 227 87
          AK149913.1 - 2 - 3 212 87
          AK150059.1 - 2 - 3 212 87
          AK150066.1 - 2 - 3 212 87
          AK154052.1 - 2 - 3 212 87
          BC023232.1 - 2 - 3 191 87
          BC065164.1 - 2 - 3 199 6829737 87
          BC066024.1 - 2 - 3 154 6829337 87
          D26091.1 - 2 - 3 215 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003472.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mcm7

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA replication
          DNA unwinding involved in DNA replication
          DNA replication initiation
          cellular response to DNA damage stimulus
          cell cycle
          cell population proliferation
          regulation of phosphorylation
          cellular response to xenobiotic stimulus
          nucleotide binding
          DNA binding
          DNA helicase activity
          single-stranded DNA binding
          helicase activity
          protein binding
          ATP binding
          hydrolase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500031665.mm.1 TC0500003472.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500031665.mm.1 NM_008568.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031665.mm.1 NM_008568.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031674.mm.1 TC0500003472.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500031674.mm.1 NM_008568.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031674.mm.1 NM_008568.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031664.mm.1 TC0500003472.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500031664.mm.1 NM_008568.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031664.mm.1 NM_008568.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031666.mm.1 TC0500003472.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500031666.mm.1 NM_008568.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031666.mm.1 NM_008568.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031677.mm.1 TC0500003472.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500031677.mm.1 NM_008568.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031677.mm.1 NM_008568.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 730579 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc