Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04207941_m1
          See other DDX3X GE Assays ›
          Gene Symbol:
          Ddx3x
          Gene Name:
          DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 3, X-linked
          Gene Aliases:
          D1Pas1-rs2, Ddx3, Fin14
          Chromosome Location:
          Chr.X: 13280496 - 13293983 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04207941_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ddx3x
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010028.3 NP_034158.1 1 - 2 142 101
          GenBank mRNA AK140990.1 - 1 - 2 143 101
          AK163250.1 - 1 - 2 117 101
          BC150862.1 - 1 - 2 96 9087773 101
          BC172016.1 - 1 - 2 96 9054302 101
          L25126.1 - 1 - 2 74 101
          Z38117.1 - 1 - 2 126 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000166.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK140990.1, AK163250.1, BC150862.1, BC172016.1, L25126.1, NM_010028.3, Z38117.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ddx3x

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          translational initiation
          regulation of translation
          apoptotic process
          chromosome segregation
          extrinsic apoptotic signaling pathway via death domain receptors
          response to virus
          regulation of gene expression
          RNA secondary structure unwinding
          positive regulation of gene expression
          negative regulation of translation
          positive regulation of cell growth
          negative regulation of cell growth
          negative regulation of protein-containing complex assembly
          DNA duplex unwinding
          stress granule assembly
          intracellular signal transduction
          ribosome biogenesis
          mature ribosome assembly
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of viral genome replication
          innate immune response
          positive regulation of translation
          positive regulation of transcription by RNA polymerase II
          positive regulation of translational initiation
          cellular response to arsenic-containing substance
          cellular response to osmotic stress
          intrinsic apoptotic signaling pathway
          positive regulation of G1/S transition of mitotic cell cycle
          negative regulation of intrinsic apoptotic signaling pathway
          nucleotide binding
          nucleic acid binding
          DNA binding
          DNA helicase activity
          RNA binding
          RNA helicase activity
          GTPase activity
          helicase activity
          protein binding
          ATP binding
          transcription factor binding
          poly(A) binding
          eukaryotic initiation factor 4E binding
          hydrolase activity
          ATP hydrolysis activity
          nucleoside-triphosphatase activity
          translation initiation factor binding
          RNA stem-loop binding
          ribosomal small subunit binding
          CTPase activity
          mRNA 5'-UTR binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00001056.mm.1 TC0X00000166.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00001056.mm.1 NM_010028.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001056.mm.1 NM_010028.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001057.mm.1 TC0X00000166.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00001057.mm.1 NM_010028.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001057.mm.1 NM_010028.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001064.mm.1 TC0X00000166.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00001064.mm.1 NM_010028.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001064.mm.1 NM_010028.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001059.mm.1 TC0X00000166.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00001059.mm.1 NM_010028.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001059.mm.1 NM_010028.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001061.mm.1 TC0X00000166.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00001061.mm.1 NM_010028.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001061.mm.1 NM_010028.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001062.mm.1 TC0X00000166.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00001062.mm.1 NM_010028.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001062.mm.1 NM_010028.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001063.mm.1 TC0X00000166.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00001063.mm.1 NM_010028.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001063.mm.1 NM_010028.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001065.mm.1 TC0X00000166.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00001065.mm.1 NM_010028.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001065.mm.1 NM_010028.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 858702 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0