Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04208181_g1
          See other SOX17 GE Assays ›
          Gene Symbol:
          Sox17
          Gene Name:
          SRY (sex determining region Y)-box 17
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 4490928 - 4497388 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04208181_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sox17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289465.1 NP_001276394.1 1 - 2 1065 114
          NM_011441.5 NP_035571.1 1 - 2 1065 114
          GenBank mRNA AK004781.1 - 1 - 2 125 114
          AK076707.1 - 1 - 2 1066 114

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001905.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK004781.1, NM_001289465.1, NM_011441.5 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sox17

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          angiogenesis
          vasculogenesis
          endoderm formation
          inner cell mass cellular morphogenesis
          heart looping
          cardiogenic plate morphogenesis
          embryonic heart tube morphogenesis
          outflow tract morphogenesis
          negative regulation of Wnt signaling pathway involved in heart development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          spermatogenesis
          gastrulation
          endoderm development
          endodermal cell fate determination
          positive regulation of gene expression
          Wnt signaling pathway
          rostrocaudal neural tube patterning
          signal transduction involved in regulation of gene expression
          negative regulation of Wnt signaling pathway
          negative regulation of cell growth
          protein destabilization
          embryonic heart tube development
          cell migration involved in gastrulation
          regulation of mesodermal cell fate specification
          negative regulation of mesodermal cell fate specification
          mRNA transcription by RNA polymerase II
          regulation of cell differentiation
          positive regulation of cell differentiation
          positive regulation of protein catabolic process
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          regulation of embryonic development
          embryonic organ development
          embryonic foregut morphogenesis
          positive regulation of skeletal muscle tissue development
          stem cell differentiation
          stem cell fate specification
          protein stabilization
          canonical Wnt signaling pathway
          endocardium formation
          cardiac cell fate determination
          endocardial cell differentiation
          common bile duct development
          gall bladder development
          endodermal digestive tract morphogenesis
          renal system development
          regulation of stem cell proliferation
          negative regulation of canonical Wnt signaling pathway
          regulation of stem cell division
          regulation of cardiac cell fate specification
          transcription cis-regulatory region binding
          DNA-binding transcription factor activity, RNA polymerase II-specific
          DNA-binding transcription activator activity, RNA polymerase II-specific
          DNA binding
          DNA-binding transcription factor activity
          transcription coactivator activity
          protein binding
          beta-catenin binding
          transcription factor binding
          sequence-specific DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100015742.mm.1 TC0100001905.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100015742.mm.1 NM_001289464.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015742.mm.1 NM_001289465.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015742.mm.1 NM_001289466.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015742.mm.1 NM_001289467.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015742.mm.1 NM_011441.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015744.mm.1 TC0100001905.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100015744.mm.1 NM_001289464.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015744.mm.1 NM_001289465.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015744.mm.1 NM_001289466.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015744.mm.1 NM_001289467.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015744.mm.1 NM_011441.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015741.mm.1 TC0100001905.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100015741.mm.1 NM_001289465.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015741.mm.1 NM_011441.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015743.mm.1 TC0100001905.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100015743.mm.1 NM_001289464.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015743.mm.1 NM_001289465.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015743.mm.1 NM_001289466.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015743.mm.1 NM_001289467.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015743.mm.1 NM_011441.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc