Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04208211_g1
          See other PRDX2 GE Assays ›
          Gene Symbol:
          Prdx2
          Gene Name:
          peroxiredoxin 2
          Gene Aliases:
          AL022839, Band-8, NkefB, PRP, PrxII, TDX1, TPx, TPx-B, TR, TSA, Tdpx1, Torin
          Chromosome Location:
          Chr.8: 84969648 - 84974317 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04208211_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prdx2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011563.5 NP_035693.3 1 - 2 118 76
          GenBank mRNA AK008433.1 - 1 - 2 123 76
          AK010653.1 - 1 - 2 93 76
          AK011963.1 - 66 76
          AK088280.1 - 1 - 2 137 76
          BC002034.1 - 1 - 2 126 3485635 76
          U20611.1 - 1 - 2 80 76
          U51679.1 - 1 - 2 55 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000974.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK008433.1, AK010653.1, AK088280.1, BC002034.1, NM_011563.5, U20611.1, U51679.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prdx2

          Gene Ontology Categories:

          Function(s) Process(es)

          respiratory burst involved in inflammatory response
          response to oxidative stress
          regulation of hydrogen peroxide metabolic process
          removal of superoxide radicals
          positive regulation of blood coagulation
          negative regulation of lipopolysaccharide-mediated signaling pathway
          negative regulation of NF-kappaB transcription factor activity
          response to lipopolysaccharide
          cellular response to oxidative stress
          T cell proliferation
          hydrogen peroxide metabolic process
          hydrogen peroxide catabolic process
          regulation of apoptotic process
          negative regulation of neuron apoptotic process
          negative regulation of T cell differentiation
          thymus development
          homeostasis of number of cells
          negative regulation of reactive oxygen species metabolic process
          negative regulation of extrinsic apoptotic signaling pathway
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          peroxidase activity
          protein binding
          thioredoxin peroxidase activity
          selenium binding
          antioxidant activity
          oxidoreductase activity
          peroxiredoxin activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800007396.mm.1 TC0800000974.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800007396.mm.1 NM_011563.6 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007395.mm.1 TC0800000974.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800007395.mm.1 NM_011563.6 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007394.mm.1 TC0800000974.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800007394.mm.1 NM_011563.6 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007402.mm.1 TC0800000974.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800007402.mm.1 NM_011563.6 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 304843 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline