Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04208414_g1
          See other MAGEB1 GE Assays ›
          Gene Symbol:
          Mageb1
          Gene Name:
          melanoma antigen, family B, 1
          Gene Aliases:
          MAGE-Xp, Mage-b1, Mage-rs1, Magel1, Smage1, dam1
          Chromosome Location:
          Chr.X: 92015191 - 92016333 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04208414_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mageb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010759.1 NP_034889.1 1 46 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00002621.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mageb1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293047 NM_010759.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293047 NM_010759.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2132932 BC119227.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2132932 NM_010759.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2132932 NM_031171.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2132932 BC145851.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2132932 BC119227.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2132932 NM_010759.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2132932 NM_031171.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2132932 BC145851.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293047 NM_010759.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293047 NM_010759.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P425815 AY099464.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P425815 BC141002.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P425815 AK019658.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P425815 AY099462.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P425815 AY099463.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293047 NM_010759.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293047 NM_010759.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122783 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32526414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32526409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32526414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14969998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32526409 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10600727 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605665 - -
          Affymetrix Mouse Gene 1.0 ST Array 10600729 - -
          Affymetrix Mouse Gene 1.0 ST Array 10600728 - -
          Affymetrix Mouse Gene 1.1 ST Array 10605664 - -
          Affymetrix Mouse Gene 1.1 ST Array 10600726 - -
          Mouse Genome 430 2.0 Array 1422345_s_at - -
          Mouse Genome 430 2.0 Array 1422345_PM_s_at - -
          Affymetrix Mouse Clariom S Array PSR0X00016376.mm.1 TC0X00002621.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016376.mm.1 NM_010759.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016376.mm.1 NM_010759.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016374.mm.1 TC0X00002621.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016374.mm.1 NM_010759.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016374.mm.1 NM_010759.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016373.mm.1 TC0X00002621.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016373.mm.1 NM_010759.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016373.mm.1 NM_010759.1 Refseq
          Mouse Expression Set 430 1422345_s_at - -
          Mouse Expression Set 430 1422345_PM_s_at - -
          Murine Genome U74v2 Set 95783_g_at - -
          Murine Genome U74v2 Set 95781_at - -
          Murine Genome U74v2 Set 95781_PM_at - -
          Murine Genome U74v2 Set 95783_PM_g_at - -
          Mouse Genome 430A 2.0 Array 1422345_s_at - -
          Mouse Genome 430A 2.0 Array 1422345_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3120411 - -
          Illumina mouse-6 v1.0 expression beadchip 3120411 - -
          Illumina mouse-6 v1.0 expression beadchip 7000184 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007000184 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003120411 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1253474 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2601802 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2601802 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253474 - -
          mouse-6 v1.1 (Illumina) scl0017145.1_232-S - -
          mouse-6 v1.1 (Illumina) scl0017145.1_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 3120411 - -
          Illumina Sentrix mouse-6.v1 bead array 7000184 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0017145.1_232-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0017145.1_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3120411 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7000184 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2601802 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2601802 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0