Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04208935_s1
          See other UFSP1 GE Assays ›
          Gene Symbol:
          Ufsp1
          Gene Name:
          UFM1-specific peptidase 1
          Gene Aliases:
          2700038N03Rik, D5Ertd655e
          Chromosome Location:
          Chr.5: 137294669 - 137295664 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04208935_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ufsp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027356.2 NP_081632.1 1 651 96
          GenBank mRNA AK012343.1 - 1 652 96
          BC016577.1 - 1 660 3584484 96
          BC087958.1 - 1 672 6751787 96

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ufsp1

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          peptidase activity
          cysteine-type peptidase activity
          hydrolase activity
          thiolester hydrolase activity
          UFM1 hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124995 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124996 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P278034 BC087958.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P278034 NM_027356.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P278034 AK012343.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P278034 BC016577.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P278034 BC087958.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P278034 NM_027356.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P278034 AK012343.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P278034 BC016577.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156422 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P278034 BC087958.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P278034 NM_027356.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P278034 AK012343.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P278034 BC016577.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P278034 BC087958.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P278034 NM_027356.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P278034 AK012343.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P278034 BC016577.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24125000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24124998 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156422 - -
          Whole Mouse Genome G4122A A_51_P278034 BC087958.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P278034 NM_027356.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P278034 AK012343.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P278034 BC016577.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P278034 BC087958.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P278034 NM_027356.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P278034 AK012343.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P278034 BC016577.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125632 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156422 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04793013 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04793019 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04793020 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04793018 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124996 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24124995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24124947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24125068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24125003 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24125000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24124995 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10526565 - -
          Affymetrix HT MG-430 PM Array Plate 1447822_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1417807_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10526564 - -
          Mouse Genome 430 2.0 Array 1447822_PM_x_at - -
          Mouse Genome 430 2.0 Array 1447822_x_at - -
          Mouse Genome 430 2.0 Array 1417807_PM_at - -
          Mouse Genome 430 2.0 Array 1417807_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70240_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70240_PM_at - -
          Mouse Expression Set 430 1417807_at - -
          Mouse Expression Set 430 1417807_PM_at - -
          Mouse Expression Set 430 1447822_PM_x_at - -
          Mouse Expression Set 430 1447822_x_at - -
          Mouse Genome 430A 2.0 Array 1417807_at - -
          Mouse Genome 430A 2.0 Array 1417807_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417807_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417807_at - -
          Affymetrix HT Mouse Genome 430B Array 1447822_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1447822_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 528915 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2680053 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002680053 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2605703 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2605703 - -
          mouse-6 v1.1 (Illumina) scl27106.1.40_71-S - -
          Illumina Sentrix mouse-6.v1 bead array 2680053 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl27106.1.40_71-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2680053 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2605703 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2605703 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl27106.1.40_71-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0