Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04212659_s1
          See other TEDDM2 GE Assays ›
          Gene Symbol:
          Teddm2
          Gene Name:
          transmembrane epididymal family member 2
          Gene Aliases:
          5830403L16Rik, AJ554213, Me9, e92
          Chromosome Location:
          Chr.1: 153849542 - 153851195 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04212659_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Teddm2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178243.3 NP_839974.2 1 651 98
          GenBank mRNA AK136655.1 - 1 653 98
          AK136685.1 - 1 652 98
          BC120491.1 - 1 605 40129414 98
          DQ402363.1 - 1 651 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003283.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Teddm2

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P85329 DQ402363.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P85329 BC120491.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P85329 BC131905.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P85329 AJ554213.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P85329 AK136655.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P85329 DQ402363.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P85329 BC120491.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P85329 BC131905.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P85329 AJ554213.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P85329 AK136655.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2148519 CR516116.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2148519 NM_178243.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 492655 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 125859 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158494 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128140 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2148519 CR516116.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2148519 NM_178243.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P85329 DQ402363.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P85329 BC120491.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P85329 BC131905.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P85329 AJ554213.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P85329 AK136655.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P85329 DQ402363.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P85329 BC120491.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P85329 BC131905.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P85329 AJ554213.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P85329 AK136655.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20728984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20728988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20728997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20728999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20728991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20728989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20728994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20728981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20728977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20728983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20728978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20729002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20728993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20728986 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128140 - -
          Whole Mouse Genome G4122A A_52_P85329 DQ402363.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P85329 BC120491.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P85329 BC131905.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P85329 AJ554213.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P85329 AK136655.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P85329 DQ402363.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P85329 BC120491.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P85329 BC131905.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P85329 AJ554213.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P85329 AK136655.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136011 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128140 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00674728 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00674719 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00173308 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00674727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20729002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20728977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00401552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00349002 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01089459 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728950 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00674732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00173309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00358702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729013 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00566669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728939 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00896950 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728956 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00939686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728965 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00348992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729001 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729002 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00674730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00338950 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00477842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00197167 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20729019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20728991 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10358923 - -
          Affymetrix Mouse Gene 1.0 ST Array 10358924 - -
          Affymetrix Mouse Gene 1.0 ST Array 10358925 - -
          Affymetrix Mouse Gene 1.0 ST Array 10358922 - -
          Affymetrix HT MG-430 PM Array Plate 1444658_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10358921 - -
          Mouse Genome 430 2.0 Array 1444658_PM_at - -
          Mouse Genome 430 2.0 Array 1444658_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 240817_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 240817_at - -
          Affymetrix Mouse Clariom S Array PSR0100026742.mm.1 TC0100003283.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100026742.mm.1 NM_178243.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100026742.mm.1 NM_178243.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100026743.mm.1 TC0100003283.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100026743.mm.1 NM_178243.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100026743.mm.1 NM_178243.3 Refseq
          Mouse Expression Set 430 1444658_PM_at - -
          Mouse Expression Set 430 1444658_at - -
          Affymetrix HT Mouse Genome 430B Array 1444658_at - -
          Affymetrix HT Mouse Genome 430B Array 1444658_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3170398 - -
          Illumina mouse-6 v1.0 expression beadchip 3170398 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003170398 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1237653 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237653 - -
          mouse-6 v1.1 (Illumina) scl0240817.1_172-S - -
          Illumina Sentrix mouse-6.v1 bead array 3170398 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0240817.1_172-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3170398 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1237653 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1237653 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0