Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04212681_m1
          See other GDPGP1 GE Assays ›
          Gene Symbol:
          Gdpgp1
          Gene Name:
          GDP-D-glucose phosphorylase 1
          Gene Aliases:
          D330012F22Rik
          Chromosome Location:
          Chr.7: 80232893 - 80241420 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04212681_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gdpgp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178752.3 NP_848867.2 1 - 2 308 97
          GenBank mRNA AK084531.1 - 1 - 2 294 97
          AK085577.1 - 314 97
          AK086166.1 - 1 - 2 310 97
          AK143704.1 - 1 - 2 334 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001147.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK084531.1, AK085577.1, AK086166.1, AK143704.1, NM_178752.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gdpgp1

          Gene Ontology Categories:

          Function(s) Process(es)

          glucose metabolic process
          nucleotide binding
          guanyl-nucleotide exchange factor activity
          transferase activity
          nucleotidyltransferase activity
          hydrolase activity
          GDP-D-glucose phosphorylase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P443544 AK143704.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P443544 BB010581.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P443544 AK084531.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P443544 AK085577.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P443544 NM_178752.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P443544 AK143704.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P443544 BB010581.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P443544 AK084531.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P443544 AK085577.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P443544 NM_178752.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986174 BC037479.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986174 AK166345.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986174 AK084531.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986174 AK085577.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986174 NM_178752.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530103 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 851647 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 899932 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928692 AK005317.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928692 AK075607.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928692 AK134856.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928692 BC017627.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928692 NM_001035509.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928692 NM_001035510.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928692 NM_025893.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 136414 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115023 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P443544 AK143704.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P443544 BB010581.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P443544 AK084531.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P443544 AK085577.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P443544 NM_178752.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P443544 AK143704.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P443544 BB010581.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P443544 AK084531.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P443544 AK085577.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P443544 NM_178752.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986174 BC037479.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986174 AK166345.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986174 AK084531.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986174 AK085577.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986174 NM_178752.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265485 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115023 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06255821 - -
          Whole Mouse Genome G4122A A_52_P443544 AK143704.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P443544 BB010581.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P443544 AK084531.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P443544 AK085577.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P443544 NM_178752.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P443544 AK143704.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P443544 BB010581.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P443544 AK084531.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P443544 AK085577.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P443544 NM_178752.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P443547 AK143704.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P443547 AK084531.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P443547 NM_178752.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137533 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115023 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06237983 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06237990 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06237975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06340928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06561618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06310143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06574115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06310138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06340930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06310139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06340933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05998696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06310150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265491 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10554395 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554393 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554394 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554396 - -
          Affymetrix HT MG-430 PM Array Plate 1444106_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10554392 - -
          Mouse Genome 430 2.0 Array 1444106_at - -
          Mouse Genome 430 2.0 Array 1444106_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 269952_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 269952_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700010050.mm.1 TC0700001147.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700010050.mm.1 NM_178752.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010050.mm.1 NM_178752.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010053.mm.1 TC0700001147.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700010053.mm.1 NM_178752.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010053.mm.1 NM_178752.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010051.mm.1 TC0700001147.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700010051.mm.1 NM_178752.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010051.mm.1 NM_178752.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010054.mm.1 TC0700001147.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700010054.mm.1 NM_178752.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010054.mm.1 NM_178752.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010052.mm.1 TC0700001147.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700010052.mm.1 NM_178752.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010052.mm.1 NM_178752.3 Refseq
          Mouse Expression Set 430 1444106_PM_at - -
          Mouse Expression Set 430 1444106_at - -
          Affymetrix HT Mouse Genome 430B Array 1444106_at - -
          Affymetrix HT Mouse Genome 430B Array 1444106_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 425404 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3060594 - -
          MouseWG-6 v2 Expression BeadChip 5270026 - -
          Illumina mouse-6 v1.0 expression beadchip 3060594 - -
          Illumina mouse-6 v1.0 expression beadchip 5270026 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003060594 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005270026 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2697043 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2697043 - -
          mouse-6 v1.1 (Illumina) scl0269952.2_132-S - -
          Illumina Sentrix mouse-6.v1 bead array 5270026 - -
          Illumina Sentrix mouse-6.v1 bead array 3060594 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00269952.1_76-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0269952.2_132-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3060594 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5270026 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1230998 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230998 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0