Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04213507_s1
          See other PPP1R3G GE Assays ›
          Gene Symbol:
          Ppp1r3g
          Gene Name:
          protein phosphatase 1, regulatory (inhibitor) subunit 3G
          Gene Aliases:
          1600032L17Rik
          Chromosome Location:
          Chr.13: 35958799 - 35970388 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04213507_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppp1r3g
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029628.1 NP_083904.1 2 1448 93
          GenBank mRNA AK005570.1 - 1 758 93
          AK050457.1 - 1 154 93
          AK079637.1 - 2 1422 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000408.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          glucose homeostasis
          positive regulation of glycogen biosynthetic process
          positive regulation of glycogen (starch) synthase activity
          protein phosphatase binding
          glycogen binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P155873 AK079637.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P155873 BQ031560.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P155873 NM_029628.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P155873 AK005570.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P155873 AK050457.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P155873 AK079637.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P155873 BQ031560.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P155873 NM_029628.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P155873 AK005570.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P155873 AK050457.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 581455 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171823 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P155873 AK079637.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P155873 BQ031560.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P155873 NM_029628.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P155873 AK005570.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P155873 AK050457.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P155873 AK079637.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P155873 BQ031560.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P155873 NM_029628.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P155873 AK005570.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P155873 AK050457.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171823 - -
          Whole Mouse Genome G4122A A_51_P155873 AK079637.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P155873 BQ031560.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P155873 NM_029628.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P155873 AK005570.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P155873 AK050457.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P155873 AK079637.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P155873 BQ031560.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P155873 NM_029628.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P155873 AK005570.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P155873 AK050457.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106428 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28946196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28946193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10543251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28946208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28946203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28946198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10803755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28946199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28946205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28946192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28946210 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10404596 - -
          Affymetrix HT MG-430 PM Array Plate 1428923_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10404595 - -
          Mouse Genome 430 2.0 Array 1428923_at - -
          Mouse Genome 430 2.0 Array 1428923_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76487_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76487_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300002468.mm.1 TC1300000408.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002468.mm.1 NM_029628.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002468.mm.1 NM_029628.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002466.mm.1 TC1300000408.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002466.mm.1 NM_029628.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002466.mm.1 NM_029628.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002467.mm.1 TC1300000408.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002467.mm.1 NM_029628.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002467.mm.1 NM_029628.1 Refseq
          Mouse Expression Set 430 1428923_PM_at - -
          Mouse Expression Set 430 1428923_at - -
          Murine Genome U74v2 Set 139276_PM_at - -
          Murine Genome U74v2 Set 139276_at - -
          Affymetrix HT Mouse Genome 430B Array 1428923_at - -
          Affymetrix HT Mouse Genome 430B Array 1428923_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 660655 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2524059 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2469081 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0