Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04235687_m1
          See other 4732416N19RIK GE Assays ›
          Gene Symbol:
          4732416N19Rik
          Gene Name:
          RIKEN cDNA 4732416N19 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 148212288 - 148235663 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04235687_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4732416N19Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015615.1 1 - 2 141 59
          GenBank mRNA AK005757.1 - 1 - 2 141 59
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY705917.1 - 1 - 2 141 Chr. 6 - 148212288 - 148235666 [+] on Build GRCm38 59

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream transmembrane and tetratricopeptide repeat containing 1 Tmtc1 Arg99,
          BC023818
          Anti-sense Chr. 6 - 148232430 - 148444845 on NCBI Build GRCm38
          Downstream ribosomal protein S4-like Rps4l 1110033J19Rik,
          AA407494,
          Rps4y2
          119.0kb Sense Chr. 6 - 148354656 - 148355596 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001752.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2418041 AK028613.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 652505 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427557 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 255221 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 305520 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 254460 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 603535 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 206095 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764290 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 316329 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 535791 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 909834 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972326 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 174613 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 48039 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166340 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06108903 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06108890 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166340 - -
          Whole Mouse Genome G4122A A_52_P604666 AK086496.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P604666 AK028613.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P413560 AK028613.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P153154 AK135908.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P153154 NM_198967.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P153154 AK033816.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P153154 AK005757.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P153154 NR_015615.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166340 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1446122_PM_at - -
          Mouse Genome 430 2.0 Array 1435261_at - -
          Mouse Genome 430 2.0 Array 1446122_PM_at - -
          Mouse Genome 430 2.0 Array 1446122_at - -
          Mouse Genome 430 2.0 Array 1435261_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600013985.mm.1 TC0600001752.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600013985.mm.1 NR_015615.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600013985.mm.1 NR_015615.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600013988.mm.1 TC0600001752.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600013988.mm.1 NR_015615.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600013988.mm.1 NR_015615.1 Refseq
          Mouse Expression Set 430 1446122_at - -
          Mouse Expression Set 430 1446122_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1446122_at - -
          Affymetrix HT Mouse Genome 430B Array 1446122_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 755293 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0