Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04236421_m1
          See other 1500011B03RIK GE Assays ›
          Gene Symbol:
          1500011B03Rik
          Gene Name:
          RIKEN cDNA 1500011B03 gene
          Gene Aliases:
          AI415201, AV141149, AW556386
          Chromosome Location:
          Chr.5: 114808196 - 114813976 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04236421_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1500011B03Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027817.1 1 - 2 193 75
          GenBank mRNA AK005199.1 - 1 - 2 141 75
          AK018264.1 - 1 - 2 142 75
          AK087533.1 - 1 - 2 134 75
          BY267441.1 - 1 - 2 194 75
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY251079.1 - 1 - 2 125 Chr. 5 - 114809037 - 114813908 [-] on Build GRCm38 75
          GenBankmRNA BY263591.1 - 1 - 2 134 Chr. 5 - 114809017 - 114813916 [-] on Build GRCm38 75
          GenBankmRNA BY271886.1 - 1 - 2 165 Chr. 5 - 114809059 - 114813948 [-] on Build GRCm38 75
          GenBankmRNA BY272840.1 - 1 - 2 194 Chr. 5 - 114809126 - 114813976 [-] on Build GRCm38 75
          GenBankmRNA BY244028.1 - 1 - 2 133 Chr. 5 - 114808991 - 114813916 [-] on Build GRCm38 75
          GenBankmRNA CX227671.1 - 1 - 2 167 Chr. 5 - 114808828 - 114813950 [-] on Build GRCm38 75
          GenBankmRNA BQ571183.1 - 1 - 2 86 Chr. 5 - 114808830 - 114813869 [-] on Build GRCm38 75
          GenBankmRNA BY277837.1 - 1 - 2 134 Chr. 5 - 114809072 - 114813916 [-] on Build GRCm38 75
          GenBankmRNA BY266792.1 - 1 - 2 134 Chr. 5 - 114809079 - 114813916 [-] on Build GRCm38 75
          GenBankmRNA CX123330.1 - 1 - 2 113 Chr. 5 - 114809122 - 114813896 [-] on Build GRCm38 75
          GenBankmRNA BY775382.1 - 1 - 2 133 Chr. 5 - 114809101 - 114813916 [-] on Build GRCm38 75
          GenBankmRNA BY287059.1 - 1 - 2 169 Chr. 5 - 114809091 - 114813951 [-] on Build GRCm38 75
          GenBankmRNA BY253552.1 - 1 - 2 134 Chr. 5 - 114809040 - 114813916 [-] on Build GRCm38 75
          GenBankmRNA DN175148.1 - 1 - 2 108 Chr. 5 - 114808838 - 114813891 [-] on Build GRCm38 75
          GenBankmRNA BY260941.1 - 1 - 2 134 Chr. 5 - 114809052 - 114813916 [-] on Build GRCm38 75
          GenBankmRNA BG088086.2 - 1 - 2 113 Chr. 5 - 114808815 - 114813896 [-] on Build GRCm38 75
          GenBankmRNA BY259859.1 - 1 - 2 131 Chr. 5 - 114809096 - 114813913 [-] on Build GRCm38 75
          GenBankmRNA CX122327.1 - 1 - 2 105 Chr. 5 - 114808919 - 114813888 [-] on Build GRCm38 75
          GenBankmRNA BB665555.1 - 1 - 2 134 Chr. 5 - 114808793 - 114813916 [-] on Build GRCm38 75
          GenBankmRNA BY279688.1 - 1 - 2 141 Chr. 5 - 114809074 - 114813923 [-] on Build GRCm38 75
          GenBankmRNA CK020247.1 - 1 - 2 140 Chr. 5 - 114808692 - 114813923 [-] on Build GRCm38 75
          GenBankmRNA CX215396.1 - 107 Chr. - - [-] on Build 75
          GenBankmRNA AI605571.1 - 1 - 2 107 Chr. 5 - 114809163 - 114813890 [-] on Build GRCm38 75
          GenBankmRNA BY287499.1 - 1 - 2 134 Chr. 5 - 114809017 - 114813916 [-] on Build GRCm38 75
          GenBankmRNA BY283680.1 - 1 - 2 143 Chr. 5 - 114809078 - 114813925 [-] on Build GRCm38 75
          GenBankmRNA BY275388.1 - 1 - 2 172 Chr. 5 - 114809098 - 114813954 [-] on Build GRCm38 76
          GenBankmRNA BY264285.1 - 1 - 2 146 Chr. 5 - 114809078 - 114813928 [-] on Build GRCm38 75
          GenBankmRNA BY245478.1 - 1 - 2 125 Chr. 5 - 114808992 - 114813908 [-] on Build GRCm38 75
          GenBankmRNA BY705378.1 - 1 - 2 141 Chr. 5 - 114808679 - 114813923 [-] on Build GRCm38 75
          GenBankmRNA BQ571060.1 - 1 - 2 127 Chr. 5 - 114808765 - 114813910 [-] on Build GRCm38 75
          GenBankmRNA BQ445468.1 - 1 - 2 117 Chr. 5 - 114808761 - 114813900 [-] on Build GRCm38 75
          GenBankmRNA BY285456.1 - 1 - 2 134 Chr. 5 - 114809036 - 114813916 [-] on Build GRCm38 75
          GenBankmRNA BY250393.1 - 1 - 2 134 Chr. 5 - 114809037 - 114813916 [-] on Build GRCm38 75
          GenBankmRNA BF449634.1 - 1 - 2 98 Chr. 5 - 114809060 - 114813881 [-] on Build GRCm38 75
          GenBankmRNA BY251129.1 - 1 - 2 134 Chr. 5 - 114809046 - 114813916 [-] on Build GRCm38 75
          GenBankmRNA CX206287.1 - 1 - 2 109 Chr. 5 - 114808662 - 114813892 [-] on Build GRCm38 75
          GenBankmRNA BY255796.1 - 1 - 2 134 Chr. 5 - 114808975 - 114813916 [-] on Build GRCm38 75
          GenBankmRNA BQ570789.1 - 1 - 2 72 Chr. 5 - 114808764 - 114813855 [-] on Build GRCm38 75
          GenBankmRNA BU524892.1 - 1 - 2 130 Chr. 5 - 114808734 - 114813910 [-] on Build GRCm38 75
          GenBankmRNA BY294272.1 - 1 - 2 134 Chr. 5 - 114809104 - 114813916 [-] on Build GRCm38 75
          GenBankmRNA AV445844.1 - 1 - 2 133 Chr. 5 - 114808967 - 114813916 [-] on Build GRCm38 75
          GenBankmRNA BY776056.1 - 1 - 2 Chr. 5 - 114809128 - 114813916 [-] on Build GRCm38 75
          GenBankmRNA BY240027.1 - 1 - 2 131 Chr. 5 - 114808998 - 114813913 [-] on Build GRCm38 75

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ankyrin repeat domain 13a Ankrd13a 1100001D10Rik,
          ANKRD13,
          AU046136
          -2.4kb Anti-sense Chr. 5 - 114774713 - 114805821 on NCBI Build GRCm38
          Downstream RIKEN cDNA 2610524H06 gene 2610524H06Rik 8.0kb Sense Chr. 5 - 114821937 - 114823468 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003734.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019577 AK161363.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019577 AK165632.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019577 BC061238.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019577 NR_027818.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407999 BU700681.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407999 AI849454.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407999 AK005199.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407999 AK018264.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407999 NR_027817.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407999 BU700681.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407999 AI849454.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407999 AK005199.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407999 AK018264.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407999 NR_027817.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481896 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 656717 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 566340 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 63243 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140575 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407999 BU700681.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407999 AI849454.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407999 AK005199.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407999 AK018264.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407999 NR_027817.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407999 BU700681.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407999 AI849454.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407999 AK005199.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407999 AK018264.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407999 NR_027817.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2019577 AK161363.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2019577 AK165632.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2019577 BC061238.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2019577 NR_027818.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01193668 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140575 - -
          Whole Mouse Genome G4122A A_51_P407999 BU700681.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407999 AI849454.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407999 AK005199.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407999 AK018264.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407999 NR_027817.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407999 BU700681.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407999 AI849454.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407999 AK005199.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407999 AK018264.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407999 NR_027817.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116411 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140575 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10532895 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532894 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532893 - -
          Affymetrix HT MG-430 PM Array Plate 1428370_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1456857_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10532892 - -
          Mouse Genome 430 2.0 Array 1456857_at - -
          Mouse Genome 430 2.0 Array 1456857_PM_at - -
          Mouse Genome 430 2.0 Array 1428370_PM_at - -
          Mouse Genome 430 2.0 Array 1428370_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66236_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66236_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500027387.mm.1 TC0500003734.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500027387.mm.1 NR_027817.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027387.mm.1 NR_027817.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027387.mm.1 NR_027818.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027387.mm.1 NR_027818.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033836.mm.1 TC0500003734.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033836.mm.1 NR_027817.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033836.mm.1 NR_027817.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027388.mm.1 TC0500003734.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500027388.mm.1 NR_027817.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027388.mm.1 NR_027817.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027388.mm.1 NR_027818.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027388.mm.1 NR_027818.1 Refseq
          Mouse Expression Set 430 1429244_PM_at - -
          Mouse Expression Set 430 1429244_at - -
          Mouse Expression Set 430 1428370_PM_at - -
          Mouse Expression Set 430 1428370_at - -
          Mouse Expression Set 430 1456857_PM_at - -
          Mouse Expression Set 430 1456857_at - -
          Mouse Genome 430A 2.0 Array 1429244_at - -
          Mouse Genome 430A 2.0 Array 1429244_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456857_at - -
          Affymetrix HT Mouse Genome 430B Array 1428370_at - -
          Affymetrix HT Mouse Genome 430B Array 1456857_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428370_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 532321 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0