Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04238030_m1
          See other 6530402F18RIK GE Assays ›
          Gene Symbol:
          6530402F18Rik
          Gene Name:
          RIKEN cDNA 6530402F18 gene
          Gene Aliases:
          mKIAA1857
          Chromosome Location:
          Chr.2: 29245116 - 29252993 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04238030_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 6530402F18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029460.1 1 - 2 527 89
          GenBank mRNA AK079298.1 - 1 - 2 515 89
          AK154396.1 - 1 - 2 456 89
          BC066084.1 - 1 - 2 527 89
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CF531942.1 - 1 - 2 525 Chr. 2 - 29248736 - 29252993 [-] on Build GRCm38 89
          GenBankmRNA BY722345.1 - 1 - 2 515 Chr. 2 - 29248728 - 29252980 [-] on Build GRCm38 89
          GenBankmRNA BY748026.1 - 1 - 2 456 Chr. 2 - 29248679 - 29252921 [-] on Build GRCm38 89
          GenBankmRNA BE862731.1 - 2 - 3 Chr. 2 - 29248203 - 29252544 [+] on Build GRCm38 89
          GenBankmRNA CD348179.1 - 1 - 2 457 Chr. 2 - 29248670 - 29252923 [-] on Build GRCm38 89
          GenBankmRNA CJ050956.1 - 1 - 2 146 Chr. 2 - 29248382 - 29252611 [-] on Build GRCm38 89

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream senataxin Setx A130090N03,
          A930037J23Rik,
          AOA2,
          AW060766,
          Als4,
          SCAR1,
          Sen1,
          mKIAA0625
          -62.6kb Anti-sense Chr. 2 - 29122588 - 29182471 on NCBI Build GRCm38
          Downstream mediator complex subunit 27 Med27 1500015J03Rik,
          2310042P07Rik,
          AA682045,
          Crsp8,
          D2Ertd434e
          93.8kb Anti-sense Chr. 2 - 29346810 - 29524790 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003245.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2248556 AK220426.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965015 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161332 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965015 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161332 - -
          Whole Mouse Genome G4122A A_51_P162340 AK018312.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P162340 AK220426.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21096126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21096140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21096159 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21096145 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21096149 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01194730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01865974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21096136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01865982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21096138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21096125 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21096131 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01194724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21096123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21096152 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21096142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21096148 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01194731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21096150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21096118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01865985 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10481365 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481359 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481360 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481363 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481361 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481358 - -
          Affymetrix HT MG-430 PM Array Plate 1441725_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1453697_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10481349 - -
          Mouse Genome 430 2.0 Array 1441725_PM_at - -
          Mouse Genome 430 2.0 Array 1453697_at - -
          Mouse Genome 430 2.0 Array 1453697_PM_at - -
          Mouse Genome 430 2.0 Array 1441725_at - -
          Affymetrix Mouse Clariom S Array PSR0200027393.mm.1 TC0200003245.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200027393.mm.1 NR_029460.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027393.mm.1 NR_029460.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027395.mm.1 TC0200003245.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200027395.mm.1 NR_029460.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027395.mm.1 NR_029460.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027398.mm.1 TC0200003245.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200027398.mm.1 NR_029460.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027398.mm.1 NR_029460.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027400.mm.1 TC0200003245.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200027400.mm.1 NR_029460.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027400.mm.1 NR_029460.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027401.mm.1 TC0200003245.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200027401.mm.1 NR_029460.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027401.mm.1 NR_029460.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027408.mm.1 TC0200003245.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200027408.mm.1 NR_029460.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027408.mm.1 NR_029460.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027396.mm.1 TC0200003245.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200027396.mm.1 NR_029460.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027396.mm.1 NR_029460.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027399.mm.1 TC0200003245.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200027399.mm.1 NR_029460.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027399.mm.1 NR_029460.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027403.mm.1 TC0200003245.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200027403.mm.1 NR_029460.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027403.mm.1 NR_029460.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027394.mm.1 TC0200003245.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200027394.mm.1 NM_133500.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027394.mm.1 NM_133501.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027394.mm.1 NR_029460.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027394.mm.1 NR_029460.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027407.mm.1 TC0200003245.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200027407.mm.1 NR_029460.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027407.mm.1 NR_029460.1 Refseq
          Mouse Expression Set 430 1450820_a_at - -
          Mouse Expression Set 430 1450820_PM_a_at - -
          Mouse Expression Set 430 1453697_at - -
          Mouse Expression Set 430 1441725_at - -
          Mouse Expression Set 430 1453697_PM_at - -
          Mouse Expression Set 430 1441725_PM_at - -
          Murine Genome U74v2 Set 137362_PM_at - -
          Murine Genome U74v2 Set 137362_at - -
          Murine Genome U74v2 Set 168922_PM_at - -
          Murine Genome U74v2 Set 168922_at - -
          Mouse Genome 430A 2.0 Array 1450820_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1450820_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1441725_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453697_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441725_at - -
          Affymetrix HT Mouse Genome 430B Array 1453697_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101740358 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0