Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04238749_s1
          See other SNORD104 GE Assays ›
          Gene Symbol:
          Snord104
          Gene Name:
          small nucleolar RNA, C/D box 104
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 106500991 - 106501063 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04238749_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Snord104
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_030703.1 1 38 70
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CJ044404.1 - 1 115 Chr. 11 - 106500915 - 106501211 [+] on Build GRCm38 70
          GenBankmRNA AA255060.1 - 1 100 Chr. 11 - 106500929 - 106501205 [+] on Build GRCm38 70
          GenBankmRNA CK620431.1 - 1 96 Chr. 11 - 106500933 - 106501211 [+] on Build GRCm38 70
          GenBankmRNA BY275986.1 - 1 111 Chr. 11 - 106500918 - 106501208 [+] on Build GRCm38 70
          GenBankmRNA AA271784.1 - 1 98 Chr. 11 - 106500931 - 106501205 [+] on Build GRCm38 70
          GenBankmRNA BY323865.1 - 1 111 Chr. 11 - 106500918 - 106501214 [+] on Build GRCm38 70
          GenBankmRNA AV459704.1 - 1 81 Chr. 11 - 106500948 - 106501211 [+] on Build GRCm38 70
          GenBankmRNA BY637987.1 - 1 118 Chr. 11 - 106500915 - 106501210 [+] on Build GRCm38 70
          GenBankmRNA W65070.1 - 102 Chr. - - [-] on Build 70
          GenBankmRNA CJ043003.1 - 1 115 Chr. 11 - 106500915 - 106501208 [+] on Build GRCm38 70
          GenBankmRNA DV056112.1 - 133 Chr. - - [-] on Build 70
          GenBankmRNA CJ325735.1 - 1 115 Chr. 11 - 106500915 - 106501214 [+] on Build GRCm38 70

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream endoplasmic reticulum (ER) to nucleus signalling 1 Ern1 9030414B18Rik,
          AI225830,
          C85377,
          Ire1a,
          Ire1alpha,
          Ire1p
          -13.2kb Anti-sense Chr. 11 - 106397620 - 106487796 on NCBI Build GRCm38
          Downstream testis expressed gene 2 Tex2 4930568E07Rik,
          AI553404,
          Def-5,
          Taz4
          1.1kb Anti-sense Chr. 11 - 106502139 - 106613418 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001783.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1440254_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10382104 - -
          Affymetrix Mouse Clariom S Array PSR1100016847.mm.1 TC1100001783.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016847.mm.1 NR_030703.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016847.mm.1 NR_030703.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 635397 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0