Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04238788_s1
          See other INAFM2 GE Assays ›
          Gene Symbol:
          Inafm2
          Gene Name:
          InaF motif containing 2
          Gene Aliases:
          5430417L22Rik, ENSMUSG00000074918
          Chromosome Location:
          Chr.2: 118745758 - 118748810 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04238788_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Inafm2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001301269.1 NP_001288198.1 1 953 97
          GenBank mRNA CX242933.1 - 1 203 97

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream phospholipase C,
          beta 2
          Plcb2 AI550384,
          B230205M18Rik,
          B230399N12
          -17.1kb Anti-sense Chr. 2 - 118707517 - 118728659 on NCBI Build GRCm38
          Downstream RIKEN cDNA A430105I19 gene A430105I19Rik Gm353 5.3kb Anti-sense Chr. 2 - 118754145 - 118762951 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2022724 CJ184953.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2022724 NR_030716.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1954521 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 387793 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 678351 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2022724 CJ184953.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2022724 NR_030716.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1954521 - -
          Whole Mouse Genome G4122A A_51_P476538 AW123519.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P476538 BB028755.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P476538 NR_030716.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P476538 AK040944.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P476538 AK077391.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129110 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21565870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21565864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01190358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21565861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21565862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21565868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21565863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21565872 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1426987_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10474807 - -
          Mouse Genome 430 2.0 Array 1426987_PM_at - -
          Mouse Genome 430 2.0 Array 1426987_at - -
          Mouse Expression Set 430 1426987_PM_at - -
          Mouse Expression Set 430 1426987_at - -
          Mouse Genome 430A 2.0 Array 1426987_PM_at - -
          Mouse Genome 430A 2.0 Array 1426987_at - -
          Affymetrix HT Mouse Genome 430A Array 1426987_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426987_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 307827 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0