Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04239510_m1
          See other ZFP133-PS GE Assays ›
          Gene Symbol:
          Zfp133-ps
          Gene Name:
          zinc finger protein 133, pseudogene
          Gene Aliases:
          OTTMUSG00000003825, Zfp133
          Chromosome Location:
          Chr.2: 144459280 - 144467794 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04239510_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp133-ps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033459.1 2 - 3 219 75
          GenBank mRNA AK137157.1 - 2 - 3 172 75
          DT931029.1 - 2 - 3 219 75
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB871509.1 - 2 - 3 163 Chr. 2 - 144459337 - 144467132 [+] on Build GRCm38 75
          GenBankmRNA BB871692.1 - 2 - 3 163 Chr. 2 - 144459337 - 144467145 [+] on Build GRCm38 75
          GenBankmRNA BB868335.1 - 2 - 3 163 Chr. 2 - 144459337 - 144467107 [+] on Build GRCm38 75
          GenBankmRNA BY722018.1 - 2 - 3 172 Chr. 2 - 144459328 - 144467374 [+] on Build GRCm38 75
          GenBankmRNA BB872896.1 - 2 - 3 163 Chr. 2 - 144459337 - 144467129 [+] on Build GRCm38 75

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cysteine and glycine-rich protein 2 binding protein Csrp2bp 2510008M08Rik,
          ATAC2,
          AU023459,
          D2Ertd473e,
          D2Wsu131e,
          E430020F17
          -56.8kb Sense Chr. 2 - 144369032 - 144407675 on NCBI Build GRCm38
          Downstream double zinc ribbon and ankyrin repeat domains 1 Dzank1 2810039F03Rik,
          6330439K17Rik,
          6430408C01,
          AI851115,
          Ankrd64
          2.8kb Anti-sense Chr. 2 - 144470555 - 144527425 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002148.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK137157.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2161675 NR_033459.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2161675 AK137157.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2161675 NR_033459.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2161675 AK137157.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04724215 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01172356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707253 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01712562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02282066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707225 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01750039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01458199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01712573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01172360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01985258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01712563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707235 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01149101 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707247 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707257 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1456238_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10476689 - -
          Affymetrix Mouse Gene 1.1 ST Array 10476680 - -
          Mouse Genome 430 2.0 Array 1456238_PM_at - -
          Mouse Genome 430 2.0 Array 1456238_at - -
          Affymetrix Mouse Clariom S Array PSR0200016931.mm.1 TC0200002148.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200016931.mm.1 NR_033459.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016931.mm.1 NR_033459.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016932.mm.1 TC0200002148.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200016932.mm.1 NR_033459.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016932.mm.1 NR_033459.1 Refseq
          Mouse Expression Set 430 1456238_PM_at - -
          Mouse Expression Set 430 1456238_at - -
          Affymetrix HT Mouse Genome 430B Array 1456238_at - -
          Affymetrix HT Mouse Genome 430B Array 1456238_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0