Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04239754_m1
          See other GM11529 GE Assays ›
          Gene Symbol:
          Gm11529
          Gene Name:
          predicted gene 11529
          Gene Aliases:
          OTTMUSG00000001775
          Chromosome Location:
          Chr.11: 96446899 - 96464547 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04239754_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm11529
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033524.1 2 - 3 507 65
          GenBank mRNA AK138406.1 - 2 - 3 508 65
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY724101.1 - 2 - 3 508 Chr. 11 - 96448323 - 96464547 [-] on Build GRCm38 65

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream homeobox B1 Hoxb1 Hox-2.9 -78.6kb Anti-sense Chr. 11 - 96365746 - 96368254 on NCBI Build GRCm38
          Downstream src family associated phosphoprotein 1 Skap1 1700091G21Rik,
          Scap1,
          Skap-55
          961.0 Anti-sense Chr. 11 - 96464576 - 96759622 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003620.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK138406.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 560850 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 414120 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 186215 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 248150 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865576 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 576140 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 614069 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 512161 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 83856 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177756 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017397 NR_033524.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017397 AK138406.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02402734 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P177756 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07452979 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P177756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09353640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09030485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09703601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09207117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09030487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09109231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09324370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09114718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09022733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09374440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09374443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09324371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09423436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09142119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09353638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09537904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09207116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09617215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09073486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09374442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09384582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09225236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09487001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09324368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28044520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09007367 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.1 ST Array 10390256 - -
          Affymetrix Mouse Clariom S Array PSR1100033163.mm.1 TC1100003620.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100033163.mm.1 NR_033524.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100033163.mm.1 NR_033524.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100033164.mm.1 TC1100003620.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100033164.mm.1 NR_033524.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100033164.mm.1 NR_033524.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0