Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04239780_s1
          See other GM16523 GE Assays ›
          Gene Symbol:
          Gm16523
          Gene Name:
          mitochondrial ribosomal protein L40 pseudogene
          Gene Aliases:
          A130054J05Rik
          Chromosome Location:
          Chr.2: 39157486 - 39162235 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04239780_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm16523
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033526.1 1 4156 95
          GenBank mRNA AK078376.1 - 1 2940 95
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BQ552004.1 - 1 Chr. 2 - 39157524 - 39158113 [+] on Build GRCm38 95

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream golgi autoantigen,
          golgin subfamily a,
          1
          Golga1 0710001G09Rik,
          2210418B03Rik,
          AW107649,
          Golgi97,
          golgin-97
          -91.9kb Sense Chr. 2 - 39016155 - 39065579 on NCBI Build GRCm38
          Downstream protein phosphatase 6,
          catalytic subunit
          Ppp6c 2310003C10Rik 32.1kb Sense Chr. 2 - 39194369 - 39226338 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003438.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2277620 AK078376.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2277620 NR_033526.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P253188 AK078376.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P253188 NR_033526.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128301 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200029503.mm.1 TC0200003438.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200029503.mm.1 NR_033526.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029505.mm.1 TC0200003438.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200029505.mm.1 NR_033526.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029504.mm.1 TC0200003438.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200029504.mm.1 NR_033526.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029507.mm.1 TC0200003438.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200029507.mm.1 NR_033526.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200029506.mm.1 TC0200003438.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200029506.mm.1 NR_033526.1 Refseq
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100580707 - -
          Illumina mouse-6 v1.0 expression beadchip 100580707 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1216609 - -
          Illumina Sentrix mouse-6.v1 bead array 100580707 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100580707 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1216609 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0