Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04239879_g1
          See other D330041H03RIK GE Assays ›
          Gene Symbol:
          D330041H03Rik
          Gene Name:
          RIKEN cDNA D330041H03 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 24409481 - 24414513 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04239879_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: D330041H03Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033554.1 5 - 6 790 104
          GenBank mRNA AK038021.1 - 903 104
          AK084778.1 - 5 - 6 685 104
          AK166073.1 - 4 - 5 562 104
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CA877678.1 - 3 - 4 358 Chr. 17 - 24410056 - 24414420 [-] on Build GRCm38 104
          GenBankmRNA BY734396.1 - 4 - 5 Chr. 17 - 24410028 - 24414510 [-] on Build GRCm38 104

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ATP-binding cassette,
          sub-family A (ABC1),
          member 3
          Abca3 1810036E22Rik,
          ABC-C,
          Abc3
          Anti-sense Chr. 17 - 24351969 - 24410204 on NCBI Build GRCm38
          Downstream ribonucleic acid binding protein S1 Rnps1 4.3kb Anti-sense Chr. 17 - 24414675 - 24425898 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001627.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK084778.1, AK166073.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P305579 AK084778.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P305579 AK166073.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P305579 NR_033554.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P305579 AK038021.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P305579 AK084778.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P305579 AK166073.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P305579 NR_033554.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P305579 AK038021.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137445 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P305579 AK084778.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P305579 AK166073.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P305579 NR_033554.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P305579 AK038021.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P305579 AK084778.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P305579 AK166073.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P305579 NR_033554.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P305579 AK038021.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P305579 AK084778.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P305579 AK166073.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P305579 NR_033554.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P305579 AK038021.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P305579 AK084778.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P305579 AK166073.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P305579 NR_033554.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P305579 AK038021.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.1 ST Array 10448559 - -
          Affymetrix Mouse Gene 1.1 ST Array 10448557 - -
          Affymetrix Mouse Clariom S Array PSR1700014403.mm.1 TC1700001627.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014403.mm.1 NR_033554.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014403.mm.1 NR_033554.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014402.mm.1 TC1700001627.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014402.mm.1 NR_033554.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014402.mm.1 NR_033554.1 Refseq
          Murine Genome U74v2 Set 135154_r_at - -
          Murine Genome U74v2 Set 135154_PM_r_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 531579 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0