Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04239969_m1
          See other BB019430 GE Assays ›
          Gene Symbol:
          BB019430
          Gene Name:
          expressed sequence BB019430
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 58696745 - 58704277 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04239969_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BB019430
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033565.1 4 - 5 326 82
          GenBank mRNA AK133046.1 - 4 - 5 327 82
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB615806.1 - 4 - 5 327 Chr. 10 - 58696745 - 58703549 [+] on Build GRCm38 82
          GenBankmRNA CF104710.1 - 4 - 5 325 Chr. 10 - 58696746 - 58703368 [+] on Build GRCm38 82

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ectodysplasin-A receptor Edar ED1R,
          ED3,
          ED5,
          EDA-A1R,
          EDA3,
          dl
          -26.1kb Anti-sense Chr. 10 - 58600780 - 58675696 on NCBI Build GRCm38
          Downstream SH3 domain containing ring finger 3 Sh3rf3 4831416G18Rik,
          Igfbp5-ip,
          Sh3md4,
          Sh3rf31
          108.1kb Sense Chr. 10 - 58812372 - 59138916 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000523.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK133046.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 467054 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 207127 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 66373 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08631854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08649732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08712340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08242836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08631864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08958761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08310168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08958759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08242830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27165062 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000003501.mm.1 TC1000000523.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000003501.mm.1 NR_033565.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003501.mm.1 NR_033565.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003499.mm.1 TC1000000523.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000003499.mm.1 NR_033565.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003499.mm.1 NR_033565.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003500.mm.1 TC1000000523.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000003500.mm.1 NR_033565.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003500.mm.1 NR_033565.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline