Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04240186_m1
          See other GM10280 GE Assays ›
          Gene Symbol:
          Gm10280
          Gene Name:
          predicted gene 10280
          Gene Aliases:
          ENSMUSG00000069883
          Chromosome Location:
          Chr.8: 113067262 - 113093334 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04240186_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm10280
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033584.1 3 - 4 441 83
          GenBank mRNA AY512935.1 - 3 - 4 441 83
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BQ560106.1 - 3 - 4 441 Chr. 8 - 113067262 - 113091933 [+] on Build GRCm38 83
          GenBankmRNA CA556331.1 - 3 - 4 441 Chr. 8 - 113067262 - 113091849 [+] on Build GRCm38 83

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream contactin associated protein-like 4 Cntnap4 Caspr4,
          E130114F09Rik
          -207.9kb Sense Chr. 8 - 112569965 - 112882707 on NCBI Build GRCm38
          Downstream RIKEN cDNA 4930571O06 gene 4930571O06Rik 423.9kb Sense Chr. 8 - 113517282 - 113550808 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001355.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AY512935.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2143105 AY512935.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2143105 NR_033584.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 476080 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 883984 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 699448 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193147 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 483528 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 473080 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 473896 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 898444 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 71543 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 45103 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109621 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06646013 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109621 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109700 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07045665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07384242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06809039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07232842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06675164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26176725 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10575660 - -
          Affymetrix Mouse Gene 1.0 ST Array 10575659 - -
          Affymetrix Mouse Gene 1.0 ST Array 10575658 - -
          Affymetrix Mouse Gene 1.1 ST Array 10575657 - -
          Affymetrix Mouse Clariom S Array PSR0800011123.mm.1 TC0800001355.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800011123.mm.1 NR_033584.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800011123.mm.1 NR_033584.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0