Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04240749_s1
          See other KIF4-PS GE Assays ›
          Gene Symbol:
          Kif4-ps
          Gene Name:
          kinesin family member 4, pseudogene
          Gene Aliases:
          4930463M05Rik, Kif4b
          Chromosome Location:
          Chr.12: 101145609 - 101149273 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04240749_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kif4-ps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033653.1 1 3355 86

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA D130020L05 gene D130020L05Rik 9430007L23Rik -56.7kb Sense Chr. 12 - 101082451 - 101088927 on NCBI Build GRCm38
          Downstream catsper channel auxiliary subunit beta Catsperb 4931410B03,
          4932415G16Rik
          255.4kb Sense Chr. 12 - 101404645 - 101626009 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001001.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_03_of_03_v2 A_68_P28686020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28686014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28686021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09762234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09762247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28686006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28686012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09762254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09762239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28686024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28686004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09762217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28686010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09762229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28686001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28686029 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10397789 - -
          Affymetrix Mouse Gene 1.1 ST Array 10397786 - -
          Affymetrix Mouse Gene 1.1 ST Array 10397788 - -
          Affymetrix Mouse Clariom S Array PSR1200007025.mm.1 TC1200001001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200007025.mm.1 NR_033653.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007025.mm.1 NR_033653.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007024.mm.1 TC1200001001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200007024.mm.1 NR_033653.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007024.mm.1 NR_033653.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0