Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04242553_s1
          See other A530053G22RIK GE Assays ›
          Gene Symbol:
          A530053G22Rik
          Gene Name:
          RIKEN cDNA A530053G22 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 60279043 - 60403707 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04242553_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A530053G22Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015565.2 2 1150 85
          GenBank mRNA AK080087.1 - 2 1151 85
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB220146.2 - 1 375 Chr. 6 - 60279043 - 60279680 [-] on Build GRCm38 85

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 5570 Gm5570 EG434019 -129.0kb Anti-sense Chr. 6 - 60078030 - 60150045 on NCBI Build GRCm38
          Downstream synuclein,
          alpha
          Snca NACP,
          alphaSYN
          451.2kb Sense Chr. 6 - 60731573 - 60829855 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998535 AK080052.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 474507 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 971581 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 588293 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 216259 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 867535 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 541893 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837870 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 35557 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 35280 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 595365 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 613465 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 775544 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 846675 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 117763 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 328670 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 27542 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928192 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 482617 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 329883 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 430309 AK042044.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 471786 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 687255 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 552185 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 864310 AK003674.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 864310 AK076498.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 864310 AK151282.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 864310 NM_026778.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 551606 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 59211 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 15604 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 496357 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 653255 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 562579 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901867 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 83760 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 511890 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901523 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 248730 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 448262 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 113035 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 803982 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 483716 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 699525 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874206 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 58393 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 29056 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33985 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 26765 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 167107 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 160089 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 135314 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 54970 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 78081 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 155453 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153422 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05915821 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05915838 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05915762 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05915843 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05915790 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01340173 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05915752 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153422 - -
          Whole Mouse Genome G4122A A_52_P137778 BC051224.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P137778 AK080052.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P137778 AK079908.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01340173 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153422 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05915752 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05915838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05872005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05875420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05867209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24488699 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10545084 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545082 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545085 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545081 - -
          Affymetrix HT MG-430 PM Array Plate 1442294_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10545079 - -
          Mouse Genome 430 2.0 Array 1442294_PM_at - -
          Mouse Genome 430 2.0 Array 1442294_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 208079_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 208079_PM_at - -
          Mouse Expression Set 430 1442294_PM_at - -
          Mouse Expression Set 430 1442294_at - -
          Affymetrix HT Mouse Genome 430B Array 1442294_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1442294_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2775763 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2775763 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2775763 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0