Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04242574_s1
          See other 1190002F15RIK GE Assays ›
          Gene Symbol:
          1190002F15Rik
          Gene Name:
          RIKEN cDNA 1190002F15 gene
          Gene Aliases:
          AI120476
          Chromosome Location:
          Chr.6: 134929092 - 134951718 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04242574_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1190002F15Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_037955.1 2 451 118
          NR_037956.1 2 476 118
          GenBank mRNA AI121041.1 - 2 412 118
          AK138072.1 - 3642 118
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BX519001.1 - 2 434 Chr. 6 - 134929155 - 134951707 [+] on Build GRCm38 118
          GenBankmRNA AI173397.1 - 2 412 Chr. 6 - 134929156 - 134951718 [+] on Build GRCm38 118

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cyclin-dependent kinase inhibitor 1B Cdkn1b AA408329,
          AI843786,
          Kip1,
          p27,
          p27Kip1
          -25.8kb Sense Chr. 6 - 134920401 - 134925525 on NCBI Build GRCm38
          Downstream apolipoprotein L domain containing 1 Apold1 Gm1075 30.3kb Sense Chr. 6 - 134982001 - 134986836 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001596.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P354373 NR_037955.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P354373 NR_037956.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P354373 AI121041.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P354373 AK004447.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P354373 AK012387.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P354373 AK012387.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P354373 NR_037955.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P354373 NR_037956.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P354373 AI121041.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P354373 AK004447.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160036 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P354373 NR_037955.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P354373 NR_037956.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P354373 AI121041.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P354373 AK004447.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P354373 AK012387.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P354373 NR_037955.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P354373 NR_037956.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P354373 AI121041.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P354373 AK004447.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P354373 AK012387.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P354373 NR_037955.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P354373 NR_037956.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P354373 AI121041.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P354373 AK004447.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P354373 AK012387.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P354373 NR_037955.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P354373 NR_037956.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P354373 AI121041.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P354373 AK004447.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P354373 AK012387.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24864390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24864344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24864386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24864375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05739553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24864379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24864393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24864404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05739554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24864412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24864387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24864378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24864345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24864348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24864407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05346575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24864408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05406607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24864342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24864381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24864380 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1441757_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600012779.mm.1 TC0600001596.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012779.mm.1 NR_037955.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012779.mm.1 NR_037955.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012779.mm.1 NR_037956.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012779.mm.1 NR_037956.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012771.mm.1 TC0600001596.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012771.mm.1 NR_037956.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012771.mm.1 NR_037956.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012766.mm.1 TC0600001596.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012766.mm.1 NR_037955.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012766.mm.1 NR_037955.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012766.mm.1 NR_037956.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012766.mm.1 NR_037956.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012768.mm.1 TC0600001596.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012768.mm.1 NR_037955.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012768.mm.1 NR_037955.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012768.mm.1 NR_037956.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012768.mm.1 NR_037956.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012776.mm.1 TC0600001596.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012776.mm.1 NR_037955.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012776.mm.1 NR_037955.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012776.mm.1 NR_037956.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012776.mm.1 NR_037956.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012778.mm.1 TC0600001596.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012778.mm.1 NR_037955.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012778.mm.1 NR_037955.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012778.mm.1 NR_037956.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012778.mm.1 NR_037956.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012775.mm.1 TC0600001596.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012775.mm.1 NR_037955.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012775.mm.1 NR_037955.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012775.mm.1 NR_037956.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012775.mm.1 NR_037956.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012770.mm.1 TC0600001596.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012770.mm.1 NR_037955.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012770.mm.1 NR_037955.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012770.mm.1 NR_037956.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012770.mm.1 NR_037956.1 Refseq
          Affymetrix HT Mouse Genome 430B Array 1441757_at - -
          Affymetrix HT Mouse Genome 430B Array 1441757_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 568643 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0