Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04242734_g1
          See other GM8909 GE Assays ›
          Gene Symbol:
          Gm8909
          Gene Name:
          predicted gene 8909
          Gene Aliases:
          EG667977, H2-Tw5l, H2-gs17
          Chromosome Location:
          Chr.17: 36141749 - 36168592 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04242734_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm8909
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081032.2 NP_001074501.2 1 - 2 93 79
          GenBank mRNA AB359231.2 - 1 - 2 93 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002004.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          antigen processing and presentation of peptide antigen via MHC class I
          signaling receptor binding
          peptide antigen binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956160 AB359231.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956160 NM_001081032.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 891589 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 3877 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956160 AB359231.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956160 NM_001081032.2 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31162851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13492190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13253477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13163846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13253483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13515919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13128854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13253474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13530626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13204095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13530622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13107013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31162787 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450718 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450722 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450720 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450721 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450719 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450717 - -
          Affymetrix Mouse Gene 1.1 ST Array 10450704 - -
          Affymetrix Mouse Clariom S Array PSR1700019229.mm.1 TC1700002004.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019229.mm.1 NM_001081032.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019229.mm.1 NM_001081032.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019231.mm.1 TC1700002004.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019231.mm.1 NM_001081032.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019231.mm.1 NM_001081032.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019233.mm.1 TC1700002004.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019233.mm.1 NM_001081032.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019233.mm.1 NM_001081032.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019235.mm.1 TC1700002004.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019235.mm.1 NM_001081032.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019235.mm.1 NM_001081032.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019226.mm.1 TC1700002004.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019226.mm.1 NM_001081032.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019226.mm.1 NM_001081032.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019234.mm.1 TC1700002004.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019234.mm.1 NM_001081032.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019234.mm.1 NM_001081032.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019238.mm.1 TC1700002004.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019238.mm.1 NM_001081032.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019238.mm.1 NM_001081032.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019228.mm.1 TC1700002004.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019228.mm.1 NM_001081032.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019228.mm.1 NM_001081032.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019240.mm.1 TC1700002004.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019240.mm.1 NM_001081032.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019240.mm.1 NM_001081032.2 Refseq
          Mouse Expression Set 430 1447260_PM_at - -
          Mouse Expression Set 430 1447260_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 675921 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2835683 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2835683 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0