Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04277727_s1
          See other E130201H02RIK GE Assays ›
          Gene Symbol:
          E130201H02Rik
          Gene Name:
          Y box protein 1 pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 120597625 - 120598475 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04277727_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: E130201H02Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_024324.1 1 623 90
          GenBank mRNA AK021400.1 - 1 625 90
          BC094539.1 - 1 598 90
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB542603.1 - 1 77 Chr. 7 - 120598176 - 120598472 [+] on Build GRCm38 90

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ATP-binding cassette,
          sub-family A (ABC1),
          member 16
          Abca16 AY243472 -52.7kb Sense Chr. 7 - 120409647 - 120544892 on NCBI Build GRCm38
          Downstream ubiquinol cytochrome c reductase core protein 2 Uqcrc2 1500004O06Rik 36.7kb Sense Chr. 7 - 120635189 - 120659523 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001729.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2066568 AK021400.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2066568 BC094539.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2066568 NR_024324.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072131 AK021400.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072131 BC094539.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072131 NR_024324.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 457613 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104621 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2066568 AK021400.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2066568 BC094539.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2066568 NR_024324.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104621 - -
          Whole Mouse Genome G4122A A_52_P261969 BU512458.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P261969 M62867.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P261969 U33196.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P261969 X57621.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P261969 BC031472.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110037 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25469244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06333239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06077702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06229565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06332221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06066154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25469295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25469237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25469242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25469307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25469269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25469239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25469266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25469273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25469267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06560714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25469276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25469280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25469236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06156540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06017674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25469294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25469271 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10556939 - -
          Affymetrix Mouse Gene 1.1 ST Array 10556938 - -
          Affymetrix Mouse Clariom S Array PSR0700014608.mm.1 TC0700001729.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700014608.mm.1 NR_024324.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700014609.mm.1 TC0700001729.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700014609.mm.1 NR_024324.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 463580 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 107000605 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2493652 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0