Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04278263_m1
          See other 2310001H17RIK GE Assays ›
          Gene Symbol:
          2310001H17Rik
          Gene Name:
          RIKEN cDNA 2310001H17 gene
          Gene Aliases:
          AI646108
          Chromosome Location:
          Chr.6: 129232623 - 129238482 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04278263_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2310001H17Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040265.1 1 - 2 304 69
          NR_040266.1 2 - 3 163 69
          NR_040268.1 2 - 3 150 69
          GenBank mRNA AK009064.1 - 1 - 2 242 69
          AV065335.2 - 1 - 2 331 69
          BE448844.1 - 1 - 2 112 69
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BE624614.1 - 1 - 2 239 Chr. 6 - 129232890 - 129237980 [-] on Build GRCm38 69
          GenBankmRNA CA457919.1 - 1 - 2 277 Chr. 6 - 129216332 - 129237982 [-] on Build GRCm38 69
          GenBankmRNA DV054834.1 - 82 Chr. - - [-] on Build 69
          GenBankmRNA W07880.1 - 162 Chr. - - [-] on Build 69
          GenBankmRNA BY708926.1 - 1 - 2 242 Chr. 6 - 129232626 - 129237982 [-] on Build GRCm38 69
          GenBankmRNA BI221897.1 - 1 - 2 246 Chr. 6 - 129232623 - 129237987 [-] on Build GRCm38 69
          GenBankmRNA DV054131.1 - 339 Chr. - - [-] on Build 69
          GenBankmRNA AA244970.1 - 1 - 2 245 Chr. 6 - 129216444 - 129237987 [-] on Build GRCm38 69
          GenBankmRNA AI006773.1 - 1 - 2 246 Chr. 6 - 129232916 - 129237987 [-] on Build GRCm38 69
          GenBankmRNA AA237611.1 - 1 - 2 302 Chr. 6 - 129232896 - 129238044 [-] on Build GRCm38 69
          GenBankmRNA AA990286.1 - 1 - 2 239 Chr. 6 - 129232863 - 129237981 [-] on Build GRCm38 69
          GenBankmRNA AA178249.1 - 1 - 2 93 Chr. 6 - 129232627 - 129237833 [-] on Build GRCm38 69
          GenBankmRNA BX518803.1 - 2 - 3 164 Chr. 6 - 129232683 - 129238482 [-] on Build GRCm38 69
          GenBankmRNA W08274.1 - 241 Chr. - - [-] on Build 69
          GenBankmRNA BE627883.1 - 2 - 3 Chr. 6 - 129232623 - 129237846 [+] on Build GRCm38 69

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream C-type lectin domain family 2,
          member d
          Clec2d Clr-b,
          Clrb,
          Ocil
          -46.5kb Anti-sense Chr. 6 - 129180615 - 129186535 on NCBI Build GRCm38
          Downstream CD69 antigen Cd69 5830438K24Rik,
          AI452015,
          AIM,
          VEA
          29.3kb Sense Chr. 6 - 129267324 - 129275369 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003227.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600025214.mm.1 TC0600003227.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025214.mm.1 NR_040265.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025214.mm.1 NR_040265.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025214.mm.1 NR_040266.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025214.mm.1 NR_040266.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025214.mm.1 NR_040268.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025214.mm.1 NR_040268.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025225.mm.1 TC0600003227.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025225.mm.1 NR_040266.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025225.mm.1 NR_040266.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025225.mm.1 NR_040268.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025225.mm.1 NR_040268.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025219.mm.1 TC0600003227.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025219.mm.1 NR_040265.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025219.mm.1 NR_040265.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025221.mm.1 TC0600003227.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025221.mm.1 NR_040265.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025221.mm.1 NR_040265.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025217.mm.1 TC0600003227.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025217.mm.1 NR_040265.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025217.mm.1 NR_040265.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025217.mm.1 NR_040266.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025217.mm.1 NR_040266.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025217.mm.1 NR_040268.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025217.mm.1 NR_040268.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025220.mm.1 TC0600003227.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025220.mm.1 NR_040265.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025220.mm.1 NR_040265.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0