Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04278297_m1
          See other A930006I01RIK GE Assays ›
          Gene Symbol:
          A930006I01Rik
          Gene Name:
          RIKEN cDNA A930006I01 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 103338601 - 103372764 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04278297_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A930006I01Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040332.1 2 - 3 276 116
          GenBank mRNA AK039421.1 - 2 - 3 241 116
          AK044304.1 - 2 - 3 211 116
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CK628534.1 - 2 - 3 182 Chr. 2 - 103340162 - 103372670 [-] on Build GRCm38 116
          GenBankmRNA BB642231.1 - 2 - 3 211 Chr. 2 - 103340133 - 103372698 [-] on Build GRCm38 116
          GenBankmRNA BG342796.1 - 276 Chr. - - [-] on Build 116
          GenBankmRNA BG298514.1 - 215 Chr. - - [-] on Build 115

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ets homologous factor Ehf 9030625L19Rik,
          AU019492
          -35.3kb Sense Chr. 2 - 103263431 - 103303275 on NCBI Build GRCm38
          Downstream cDNA sequence BC016548 BC016548 35.6kb Sense Chr. 2 - 103377643 - 103390187 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004225.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2358216 AK044304.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 860120 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765391 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 573771 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 175172 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 203011 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 397183 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 37046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 211602 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 856931 AK041267.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 113981 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 669445 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 190601 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 89714 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 167069 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155922 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2358216 AK044304.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04629087 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04629074 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155922 - -
          Whole Mouse Genome G4122A A_52_P502181 AK039421.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P502181 AK044304.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125030 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01212513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01895452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01212509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01656156 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01656158 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01839528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01915699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01192665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01915703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01339574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01262898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01839527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01424383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01192825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01374585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01376641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02204256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01768070 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01591165 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01601027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01339565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02204254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01656149 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01339561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01656152 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482771 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01212508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01335539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01192667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01656153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482887 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482873 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01819540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482866 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01262912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01339571 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482824 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01339566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482705 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01950594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01860019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01846016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21482761 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200035472.mm.1 TC0200004225.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200035472.mm.1 NR_040332.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035472.mm.1 NR_040332.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 619806 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100510451 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0