Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04278300_m1
          See other A330093E20RIK GE Assays ›
          Gene Symbol:
          A330093E20Rik
          Gene Name:
          RIKEN cDNA A330093E20 gene
          Gene Aliases:
          AU043608
          Chromosome Location:
          Chr.18: 45683487 - 46045260 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04278300_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A330093E20Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040342.1 3 - 4 329 59
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB641117.1 - 3 - 4 331 Chr. 18 - 45685193 - 46045260 [-] on Build GRCm38 59
          GenBankmRNA CK332073.1 - 2 - 3 139 Chr. 18 - 45685130 - 45854544 [-] on Build GRCm38 59
          GenBankmRNA BG084391.2 - 2 - 3 139 Chr. 18 - 45685354 - 45854544 [-] on Build GRCm38 59

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          31706
          Gm31706 -542.3kb Anti-sense Chr. 18 - 45044811 - 45141138 on NCBI Build GRCm38
          Downstream tripartite motif-containing 36 Trim36 D18Wsu100e,
          haprin
          328.2kb Sense Chr. 18 - 46165300 - 46212607 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001288.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC1800001374.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2269154 AK039712.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 518204 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 67001 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 88795 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 896819 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 541877 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 236981 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 264681 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 775766 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582553 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837346 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 673492 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 278762 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 181375 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 636713 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 877947 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 84394 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 835449 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 438036 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 707391 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 693470 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544153 AK142225.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544153 AK173097.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544153 AY481619.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544153 AY481620.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544153 AY481621.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544153 AY481622.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544153 AY481623.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544153 BC031201.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544153 BC057352.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544153 BC105578.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544153 NM_001001882.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 196840 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 950571 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 828247 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 561029 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 747628 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 440354 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487968 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 836546 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914798 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 794283 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 162291 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 830108 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701029 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 663076 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 644483 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 326925 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847558 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481101 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 684494 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 17960 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 600558 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906915 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 493672 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 168442 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 877392 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 381661 AK047264.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 381661 AK140603.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 329539 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 821635 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 799126 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 829934 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 725577 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 678837 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 774491 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 31502 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290400 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 834479 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 539474 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567691 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 514950 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 361474 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 148421 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 252988 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 14922 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 738489 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 124404 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 484234 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33121 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 29784 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 28467 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 126153 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 26905 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 174947 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 23811 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142423 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 47057 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 3642 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 118073 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 42237 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 133339 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114140 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2269154 AK039712.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08987090 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03465801 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08987206 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08987147 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08986948 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08987258 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08987124 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114140 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08986997 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03465768 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08987168 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08986916 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08987046 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08987238 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03465992 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03465787 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08987190 - -
          Whole Mouse Genome G4122A A_52_P342052 AK039712.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137305 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08987206 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08987090 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08987168 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08987238 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114140 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08986916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31673368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674256 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13678106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31673370 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14036056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13857285 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13950083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13678116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13731941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31673366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13751839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13900696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31673372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13644741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13615864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13643676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14131521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13666961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14016926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674248 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14131524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13837753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31673359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13900697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31673373 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14067382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13678097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31673364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13666943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13906525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14086133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14067380 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31673365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13864038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13900693 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13644746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31673361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674245 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31673375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31673363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13731948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31674032 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1800009567.mm.1 TC1800001288.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009567.mm.1 NR_040342.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009567.mm.1 NR_040342.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009568.mm.1 TC1800001288.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009568.mm.1 NR_040342.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009568.mm.1 NR_040342.1 Refseq
          Murine Genome U74v2 Set 129102_PM_at - -
          Murine Genome U74v2 Set 129102_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 407939 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc